145262
(Ethoxycarbonylmethyl)dimethylsulfonium bromide
97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
97%
form
solid
mp
90-94 °C (lit.)
functional group
ester
thioether
SMILES string
Br.CCOC(=O)C[S](C)C
InChI
1S/C6H13O2S.BrH/c1-4-8-6(7)5-9(2)3;/h4-5H2,1-3H3;1H/q+1;/p-1
InChI key
JXFPTJYKYKVENJ-UHFFFAOYSA-M
Application
(Ethoxycarbonylmethyl)dimethylsulfonium bromide was used in synthesis of α-sulfanyl-β-amino acid derivatives by using nanocrystalline magnesium oxide. It was used during cyclopropanation of (-)-(S)-2-(p-tolylsulfinyl)cyclopent-2-enone.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Constrained cycloalkyl analogues of glutamic acid: stereocontrolled synthesis of (+)-2-aminobicyclo [3.1. 0] hexane-2, 6-dicarboxylic acid (LY354740) and its 6-phosphonic acid analogue
Tetrahedron Asymmetry, 21(11), 1486-1493 (2010)
Synthesis of a-sulfanyl-?-amino acid derivatives by using nanocrystalline magnesium oxide.
Tetrahedron, 66(27), 5042-5052 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service