Skip to Content
Merck
All Photos(3)

Key Documents

143618

Sigma-Aldrich

2,5-Dimethyl-2,5-hexanediol

97%

Synonym(s):

1,1,4,4-Tetramethyl-1,4-butanediol, 2,5-Dihydroxy-2,5-dimethylhexane, 2,5-Dimethyl-2,5-dihydroxyhexane, 2,5-Dimethyl-2,5-hexanediol, 2,5-Dimethylhexane-2,5-diol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(OH)CH2CH2C(OH)(CH3)2
CAS Number:
Molecular Weight:
146.23
Beilstein:
1361437
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

214-215 °C (lit.)

mp

86-90 °C (lit.)

functional group

hydroxyl

SMILES string

CC(C)(O)CCC(C)(C)O

InChI

1S/C8H18O2/c1-7(2,9)5-6-8(3,4)10/h9-10H,5-6H2,1-4H3

InChI key

ZWNMRZQYWRLGMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,5-Dimethyl-2,5-hexanediol on heteropoly acid catalyzed dehydration yields cyclic ethers via stereospecific intramolecular SN2 mechanism. It reacts with nitriles in concentrated sulfuric acid to yield Δ1-pyrrolines.

Application

2,5-Dimethyl-2,5-hexanediol was used in the synthesis of six- and seven-membered heterocyclic boron compounds containing intramolecular N-B bond.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Transformation of diols in the presence of heteropoly acids under homogeneous and heterogeneous conditions.
Torok B, et al.
J. Mol. Catal. A: Chem., 107(1), 305-311 (1996)
Synthesis and Spectroscopic Characterization of the First Mixed Six-and Seven-Membered Heterocyclic Boron Compounds With Intramolecular N? B Bond.
Goyal S, et al.
Main Group Metal Chemistry, 32(2), 55-64 (2009)
Nitriles in Nuclear Heterocyclic Synthesis. II.
MEYERS AI and RITTER JJ.
The Journal of Organic Chemistry, 23(12), 1918-1922 (1958)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service