Skip to Content
Merck
All Photos(1)

Key Documents

142352

Sigma-Aldrich

4-Cyanopyridine N-oxide

96%

Synonym(s):

Isonicotinonitrile 1-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4N2O
CAS Number:
Molecular Weight:
120.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

223-225 °C (lit.)

solubility

1 M NH4OH: soluble 5 mg/mL, clear, colorless (methanol)

SMILES string

[O-][n+]1ccc(cc1)C#N

InChI

1S/C6H4N2O/c7-5-6-1-3-8(9)4-2-6/h1-4H

InChI key

QNCSFBSIWVBTHE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carl M Blumenfeld et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 18(9), 1091-1100 (2017-01-18)
Globally scalable sunlight-driven devices that convert solar energy into storable fuels will require efficient light absorbers that are made of non-precious elements. Suitable photoanode materials are yet to be discovered. Here we utilised the timesaving nature of pulsed-laser-in-liquids synthesis and
Construction of Two Coordination Complexes Based on 5-(4-Pyridine N-oxide) Tetrazole (4-ptz) Generated by an in-situ [2+ 3] Cycloaddition Reaction.
Xiang J, et al.
Zeitschrift fur Anorganische und Allgemeine Chemie, 639(3-4), 563-568 (2013)
Zhao Xiang Yu et al.
Acta crystallographica. Section C, Crystal structure communications, 60(Pt 4), m194-m196 (2004-04-09)
The intermediate [Cd(4-PTZ)(2)(H(2)O)(4)] [4-PTZ is 5-(4-pyridyl N-oxide)tetrazolate, C(6)H(4)N(5)O], (I), in the synthesis of 4-HPTZ, (II), from the cycloaddition reaction of 4-cyanopyridine N-oxide with NaN(3) in water using CdCl(2) as catalyst, was structurally characterized. The unique Cd atom lies on a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service