Skip to Content
Merck
All Photos(2)

Key Documents

135690

Sigma-Aldrich

3,5-Diisopropyl-2-hydroxybenzoic acid

98%

Synonym(s):

3,5-Diisopropylsalicylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2C6H2-2-(OH)CO2H
CAS Number:
Molecular Weight:
222.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

113-115 °C (lit.)

functional group

carboxylic acid

SMILES string

CC(C)c1cc(C(C)C)c(O)c(c1)C(O)=O

InChI

1S/C13H18O3/c1-7(2)9-5-10(8(3)4)12(14)11(6-9)13(15)16/h5-8,14H,1-4H3,(H,15,16)

InChI key

XUFUYOGWFZSHGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,5-Diisopropyl-2-hydroxybenzoic acid plays the role of OH-inactivating ligand in its complexes with copper(II).

Application

3,5-Diisopropyl-2-hydroxybenzoic acid was used as starting reagent in the synthesis of Zn(II) and Cd(II) carboxylate complexes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monika Homoncik et al.
Blood coagulation & fibrinolysis : an international journal in haemostasis and thrombosis, 14(2), 131-138 (2003-03-13)
We examined the influence of 3,5-diisopropylsalicylic acid (3,5-DIPS) and calcium(II)3 (3,5-diisopropylsalicylate)6 (H2 O)6 [Ca(II)3 (3,5-DIPS)6 ], a new activator of calcium-dependent calmodulin-triggered nitric oxide synthase, on thrombin-induced platelet P-selectin expression. Citrated whole blood samples were incubated with either ethanol vehicle
Eugene O Apostolov et al.
Radiation research, 172(4), 481-492 (2009-09-24)
Gamma radiation is known to induce cell death in several organs. This damage is associated with endonuclease-mediated DNA fragmentation; however, the enzyme that produces the latter and is likely to cause cell death is unknown. To determine whether the most
V Brumas et al.
Journal of inorganic biochemistry, 101(4), 565-577 (2007-01-20)
The purpose of this study was to identify the low molecular mass complexes formed between copper(II) and 3,5-diisopropylsalicylic acid (Dips) in physiological conditions. Copper(II)-Dips complex equilibria were determined using glass electrode potentiometry and their solution structures checked by UV-visible (UV-vis)
Michèle Lamoureux et al.
Journal of fluorescence, 20(3), 631-643 (2010-04-07)
The coordination complexes (DIP)(2)Ru(CH(3)bpyCOOH) and (DIP)(2)Ru(COOHbpyCOOH), where DIP and bpy are diphenylphenanthroline and bispyridine, have been recently proposed as fluorescent markers of nuclear DNA (Musatkina et al., J. Inorg. Biochem. 101:1086-1089, 2007), but no DNA binding investigation and no quantitative
L F Charles et al.
Journal of materials science. Materials in medicine, 21(6), 1845-1854 (2010-03-20)
Currently, the bone-repair biomaterials market is dominated by high modulus metals and their alloys. The problem of stress-shielding, which results from elastic modulus mismatch between these metallic materials and natural bone, has stimulated increasing research into the development of polymer-ceramic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service