Skip to Content
Merck
All Photos(2)

Key Documents

134910

Sigma-Aldrich

(1R,2S)-(−)-Ephedrine

98%

Synonym(s):

(−)-Ephedrine, (1R,2S)-(−)-α-(1-Methylaminoethyl)benzyl alcohol, (1R,2S)-(−)-2-Methylamino-1-phenyl-1-propanol, L-α-(1-Methylaminoethyl)benzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH[CH(NHCH3)CH3]OH
CAS Number:
Molecular Weight:
165.23
Beilstein:
2208730
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

optical activity

[α]21/D −41°, c = 5 in 1 M HCl

bp

255 °C (lit.)

mp

37-39 °C (lit.)

density

1.124 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl
phenyl

storage temp.

2-8°C

SMILES string

CN[C@@H](C)[C@H](O)c1ccccc1

InChI

1S/C10H15NO/c1-8(11-2)10(12)9-6-4-3-5-7-9/h3-8,10-12H,1-2H3/t8-,10-/m0/s1

InChI key

KWGRBVOPPLSCSI-WPRPVWTQSA-N

General description

(1R,2S)-(-)-Ephedrine, a chiral β-amino alcohol, is commonly used as a chiral auxiliary in asymmetric synthesis. It undergoes reduction to form (+)-methamphetamine.

Application

(1R,2S)-(-)-Ephedrine can be used in the preparation of optically active phosphine-phosphonite ligands and chirally-substituted cyclopentadienyl (Cp) ligands. It reacts with phosphorus trichloride to form (2R,4S,5R)-2-chloro-3,4-dimethyl-5-phenyl-1,3,2-oxazaphospholidine. It can also act as a chiral resolving agent for (±)-mandelic acid.
Versatile chiral synthon, employed in catalysis and in the preparation of optically pure sulfoxides and oxazolidines.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A novel nucleoside phosphoramidite synthon derived from 1R, 2S-ephedrine
Iyer RP, et al
Tetrahedron Asymmetry, 6(5), 1051-1054 (1995)
A L Carey et al.
Diabetologia, 56(1), 147-155 (2012-10-16)
Brown adipose tissue (BAT) activation increases energy consumption and may help in the treatment of obesity. Cold exposure is the main physiological stimulus for BAT thermogenesis and the sympathetic nervous system, which innervates BAT, is essential in this process. However
Synthetic Communications, 829-829 (1992)
A novel chiral cyclopentadienyl ligand based on ephedrine
van der Zeijden AAH
Tetrahedron Asymmetry, 6(4), 913-918 (1995)
(1R,2S)-Ephedrine
Soai K and Hitchcock SR
e-EROS Encyclopedia of Reagents for Organic Synthesis. null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service