Skip to Content
Merck
All Photos(3)

Key Documents

134805

Sigma-Aldrich

Melissic acid

≥98%

Synonym(s):

Triacontanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)28COOH
CAS Number:
Molecular Weight:
452.80
Beilstein:
1803689
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

powder

mp

92-94 °C (lit.)

solubility

chloroform: soluble 25 mg/mL, clear, colorless to faintly yellow

functional group

carboxylic acid

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C30H60O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29-30(31)32/h2-29H2,1H3,(H,31,32)

InChI key

VHOCUJPBKOZGJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Melissic acid is isolated from the pericarp of Trichosanthes rosthornii.

Application

Melissic acid was used in a study on identification and determination of very-long-chain polyunsaturated fatty acid in freshwater crustacean species Bathynella natans.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Studies on the Chemical Constituents of the Pericarp of Trichosanthes rosthornii Harms [J].
Zhimao C and Jingming L.
Zhongguo Zhong Yao Za Zhi = Zhongguo zhongYao ZaZhi = China Journal of Chinese Materia Medica, 2, 15-15 (1991)
Tetratriacontanonaenoic acid, first natural acid with nine double bonds isolated from a crustacean Bathynella natans.
Rezanka T and Dembitsky VM.
Tetrahedron, 60(19), 4261-4264 (2004)
Tomonobu Uchino et al.
Cancer chemotherapy and pharmacology, 86(2), 233-243 (2020-07-16)
Erlotinib, an inhibitor of the epidermal growth factor receptor tyrosine kinase, causes skin disorders such as dry skin, which impairs the skin barrier function. Stratum corneum (SC) lipids play an important role in skin barrier function; therefore, this study aimed
Maria Winterfeld et al.
Nature communications, 9(1), 3666-3666 (2018-09-12)
The mobilization of glacial permafrost carbon during the last glacial-interglacial transition has been suggested by indirect evidence to be an additional and significant source of greenhouse gases to the atmosphere, especially at times of rapid sea-level rise. Here we present
Amy C F King et al.
Talanta, 194, 233-242 (2019-01-06)
The majority of atmospheric compounds measured in ice cores are inorganic, while analysis of their organic counterparts is a less well developed field. In recent years, understanding of formation, transport pathways and preservation of these compounds in ice and snow

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service