Skip to Content
Merck
All Photos(1)

Documents

132632

Sigma-Aldrich

3,5-Dimethoxyphenol

99%

Synonym(s):

Phloroglucinol dimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3OH
CAS Number:
Molecular Weight:
154.16
Beilstein:
1239156
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

172-175 °C/17 mmHg (lit.)

mp

40-43 °C (lit.)

SMILES string

COc1cc(O)cc(OC)c1

InChI

1S/C8H10O3/c1-10-7-3-6(9)4-8(5-7)11-2/h3-5,9H,1-2H3

InChI key

XQDNFAMOIPNVES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

3,5-Dimethoxyphenol has been used in a study on iron porphyrin phenoxides. It was used to develop a new analytical method using LC-MS that enables the simultaneous identification and quantification of new alkaloids and the alkaloidal diterpenoids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iron porphyrin phenoxides: models for some hemoglobin mutants.
Ainscough E_W, et al.
Journal of the American Chemical Society, 100(24), 7585-7591 (1978)
Modern analytical procedures for the determination of taxus alkaloids in biological material.
F Musshoff et al.
International journal of legal medicine, 122(4), 357-358 (2008-05-07)
J Stríbrný et al.
Soudni lekarstvi, 55(3), 36-39 (2010-10-15)
Autopsy findings of fatal intoxication with yew (Taxus baccata) are nonspecific. A presence of plant residues in the digestive tract can signalize yew intoxication. If yew decoction is consumed, plant residues are not found. In such a case the intoxication
Márcia Pessêgo et al.
Journal of pharmacy & bioallied sciences, 3(1), 128-134 (2011-03-25)
Nitrosative deamination of DNA bases induced by reaction with reactive nitrogen species (RNS) has been pointed out as a probable cause of mutagenesis. (Poly)phenols, present in many food items from the Mediterranean diet, are believed to possess antinitrosating properties due
R Froldi et al.
Journal of analytical toxicology, 34(1), 53-56 (2010-01-30)
Taxus baccata is a widely distributed yew often associated with cases of fatal intoxication, which is related to the high amounts of cardiotoxic alkaloids, taxine A and taxine B, contained in its leaves. In this paper, a case of Taxus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service