Skip to Content
Merck
All Photos(1)

Documents

129100

Sigma-Aldrich

Ethyl 5-bromovalerate

98% (contains silver wool as stabilizer)

Synonym(s):

5-Bromopentanoic acid ethyl ester, Ethyl 5-bromopentanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)4COOC2H5
CAS Number:
Molecular Weight:
209.08
Beilstein:
1752023
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98% (contains silver wool as stabilizer)

form

liquid

contains

(Contains silver wool as stabilizer)

refractive index

n20/D 1.458 (lit.)

bp

104-109 °C/12 mmHg (lit.)

density

1.321 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CCCCBr

InChI

1S/C7H13BrO2/c1-2-10-7(9)5-3-4-6-8/h2-6H2,1H3

InChI key

AFRWBGJRWRHQOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl 5-bromovalerate was used in the preparation of S(γ-Carboxypropyl)-DL-homocysteine and S(δ-Carboxybutyl)-DL-homocysteine. It was also used in the preparation of ethyl 5-azidovalerate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1,3-Dipolar cycloaddition of azides with electron-deficient alkynes under mild condition in water.
Zengmin Li, et al.
Tetrahedron Letters, 45(15), 3143-3146 (2004)
W M Awad et al.
The Journal of biological chemistry, 258(21), 12790-12792 (1983-11-10)
We described recently the purification and preliminary characterization of human hepatic betaine: homocysteins S-methyltransferase (Skiba, W. E., Taylor, M. P., Wells, M. S., Mangum, J. H., and Awad, W. M., Jr. (1982) J. Biol. Chem. 258, 14944-14948) where it was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service