Skip to Content
Merck
All Photos(1)

Key Documents

128244

Sigma-Aldrich

Ethyl diethoxyacetate

97%

Synonym(s):

Glyoxylic acid ethyl ester diethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2CHCOOC2H5
CAS Number:
Molecular Weight:
176.21
Beilstein:
878579
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.410 (lit.)

bp

199 °C (lit.)

density

0.985 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(OCC)C(=O)OCC

InChI

1S/C8H16O4/c1-4-10-7(9)8(11-5-2)12-6-3/h8H,4-6H2,1-3H3

InChI key

XCLBIKIQSCTANZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl diethoxyacetate was used in the preparation of a fatty acid analog containing DTS, 12,13-dioxotetradecanoic acid di(N-methyl-thiosemicarbazone) (FA-DTS).

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Arano et al.
International journal of radiation applications and instrumentation. Part A, Applied radiation and isotopes, 40(9), 745-749 (1989-01-01)
For the development of 62Cu labeled fatty acid analogs, two fatty acid analogs, containing dithiosemicarbazone (DTS) molecule as the 62Cu coordinating site, were designed and synthesized: a fatty acid analog containing DTS molecule at the omega-position, (a) the 12,13-dioxotetradecanoic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service