Skip to Content
Merck
All Photos(1)

Key Documents

117196

Sigma-Aldrich

(2-Bromoethyl)trimethylammonium bromide

98%

Synonym(s):

Bromocholine Bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2N(CH3)3Br
CAS Number:
Molecular Weight:
246.97
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

244 °C (dec.) (lit.)

SMILES string

[Br-].C[N+](C)(C)CCBr

InChI

1S/C5H13BrN.BrH/c1-7(2,3)5-4-6;/h4-5H2,1-3H3;1H/q+1;/p-1

InChI key

OINMNSFDYTYXEQ-UHFFFAOYSA-M

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haibo Wang et al.
Nature structural & molecular biology, 27(1), 8-13 (2019-12-11)
Recognition of histone-modified nucleosomes by specific reader domains underlies the regulation of chromatin-associated processes. Whereas structural studies revealed how reader domains bind modified histone peptides, it is unclear how reader domains interact with modified nucleosomes. Here, we report the cryo-electron
Caitlin I Stoddard et al.
Molecular cell, 73(1), 73-83 (2018-11-13)
DNA methylation and H3K9me are hallmarks of heterochromatin in plants and mammals, and are successfully maintained across generations. The biochemical and structural basis for this maintenance is poorly understood. The maintenance DNA methyltransferase from Zea mays, ZMET2, recognizes dimethylation of
Francesca Munari et al.
PloS one, 8(4), e60887-e60887 (2013-04-16)
As essential components of the molecular machine assembling heterochromatin in eukaryotes, HP1 (Heterochromatin Protein 1) proteins are key regulators of genome function. While several high-resolution structures of the two globular regions of HP1, chromo and chromoshadow domains, in their free
Phillip A Dumesic et al.
Molecular cell, 79(1), 127-139 (2020-05-22)
C.neoformans Dnmt5 is an unusually specific maintenance-type CpG methyltransferase (DNMT) that mediates long-term epigenome evolution. It harbors a DNMT domain and SNF2 ATPase domain. We find that the SNF2 domain couples substrate specificity to an ATPase step essential for DNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service