Skip to Content
Merck
All Photos(1)

Key Documents

10803

Sigma-Aldrich

Apiezon® N

grease (consistency similar to rubber)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
NACRES:
NA.22
form:
grease (consistency similar to rubber)

form

grease (consistency similar to rubber)

Quality Level

impurities

polymer hydrocarbon with higher molecular weight

General description

Apiezon®N enhances the thermal contact of the sample during thermal conductivity measurements in single-wall carbon nanotube epoxy composites. It was coated on single crystals of LiFeAs to provide protection.

Legal Information

Apiezon is a registered trademark of M & I Materials Ltd.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal conductivity and interfacial resistance in single-wall carbon nanotube epoxy composites.
Bryning MB, et al.
Applied Physics Letters, 87(16), 161909-161909 (2005)
Anisotropic upper critical field and possible Fulde-Ferrel-Larkin-Ovchinnikov state in the stoichiometric pnictide superconductor LiFeAs.
Cho K, et al.
Physical Review. B, Condensed Matter and Materials Physics, 83(6), 060502-060502 (2011)
Linda D Stewart et al.
BMC veterinary research, 13(1), 131-131 (2017-05-14)
The European badger is an important wildlife reservoir of Mycobacterium bovis implicated in the spread of bovine tuberculosis in the United Kingdom and Ireland. Infected badgers are known to shed M. bovis in their urine and faeces, which may contaminate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service