Skip to Content
Merck
All Photos(1)

Key Documents

107891

Sigma-Aldrich

2-Isopropoxyethanol

99%

Synonym(s):

Ethylene glycol monoisopropyl ether, Isopropylglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOCH2CH2OH
CAS Number:
Molecular Weight:
104.15
Beilstein:
1732184
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39020731
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

refractive index

n20/D 1.41 (lit.)

bp

42-44 °C/13 mmHg (lit.)

density

0.903 g/mL at 25 °C (lit.)

functional group

ether
hydroxyl

SMILES string

CC(C)OCCO

InChI

1S/C5H12O2/c1-5(2)7-4-3-6/h5-6H,3-4H2,1-2H3

InChI key

HCGFUIQPSOCUHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Isopropoxyethanol molecules self-associate by hydrogen bonding between OH group and partially etheric O atom and also by hydrophobic interaction between isopropoxyethyl groups.

Application

2-Isopropoxyethanol was used to study the products of the gas-phase reactions of OH radical with n-butyl methyl ether and 2-isopropoxyethanol in the presence of NO. 2-Isopropoxyethanol was used to develop methodology for evaluating electromagnetic sensor performance in nonrelaxing and nonconducting (e.g., sandy soil) media.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

109.4 - 113.0 °F - Equilibrium method

Flash Point(C)

43 - 45 °C - Equilibrium method

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermodynamic properties of aqueous solution of 2-isopropoxyethanol at 25? C.
Tamura K, et al.
Journal of Solution Chemistry, 28(6), 777-789 (1999)
Products of the gas-phase reactions of the OH radical with n-butyl methyl ether and 2-isopropoxyethanol: Reactions of ROC (?) < radicals.
Aschmann SM and Atkinson R.
International Journal of Chemical Kinetics, 31(7), 777-789 (1999)
Andrzej Starek et al.
International journal of occupational medicine and environmental health, 17(3), 339-346 (2005-02-03)
Administration of ethylene glycol monoalkyl ethers to rodents causes acute hemolytic anemia. Metabolic activation of these chemicals to alkoxyacetic acids is required to develop hemolytic effect. Current study was undertaken to compare the hemolytic activity of isopropoxyethanol (IPE) and phenoxyethanol
Standardizing characterization of electromagnetic water content sensors.
Jones SB, et al.
Vadose Zone Journal, 4(4), 1048-1058 (2005)
F Welsch et al.
Environmental health perspectives, 57, 125-133 (1984-08-01)
Cell-cell communication, possibly through gap junctions, is a fundamental event for the differentiation of embryonal tissues. Chemical substances which can interfere with this process may be able to disrupt embryogenesis. We have examined the response of a normal diploid human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service