Skip to Content
Merck
All Photos(3)

Key Documents

102091

Sigma-Aldrich

3-Diethylaminophenol

97%

Synonym(s):

3-(Diethylamino)phenol, m-(Diethylamino)phenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NC6H4OH
CAS Number:
Molecular Weight:
165.23
Beilstein:
908212
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

bp

170 °C/15 mmHg (lit.)

mp

69-72 °C (lit.)

functional group

amine

SMILES string

CCN(CC)c1cccc(O)c1

InChI

1S/C10H15NO/c1-3-11(4-2)9-6-5-7-10(12)8-9/h5-8,12H,3-4H2,1-2H3

InChI key

WAVOOWVINKGEHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Diethylaminophenol is raw material required for the synthesis of Rhodamine derivatives.

Application

3-Diethylaminophenol was used as starting reagent for the synthesis of silyloxyaniline. It was used in the synthesis of Nile Red and its hydroxy-derivative.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

285.8 °F

Flash Point(C)

141 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Novel Synthesis of Rhodamine Derivatives.
Zhi-jian C and Xiao-jun P.
Liaoning Chemical Industry, 6 (2004)
Photoconductive Nile red cyclopalladated metallomesogens.
Ionescu A, et al.
Journal of Materials Chemistry, 22(44), 23617-23626 (2012)
Vanessa Jesus Ferreira et al.
Analytical methods : advancing methods and applications, 13(2), 267-273 (2020-12-29)
A method based on ultrasound-assisted emulsification liquid-liquid microextraction (USAEME) for cadmium determination by flame atomic absorption spectrometry (FAAS) was developed in this work. USAEME is based on the use of the mixture of 1,2-dichloroethane and trichloroethylene as an acceptor phase
Analú P Santos et al.
Journal of the science of food and agriculture, 99(10), 4609-4614 (2019-03-21)
In the present work, a simple and rapid method was proposed for the preconcentration of cadmium using ultrasound-assisted temperature-controlled ionic liquid microextraction (TC-IL-LPME). The dispersion of the ionic liquid (1-Hmim[PF6 ]) in the aqueous phase was performed by heating it
Xinyu Tian et al.
ChemPlusChem, 85(8), 1639-1645 (2020-08-05)
We report the synthesis, characterization, and photophysical and ion-binding properties of deep-red to near-infrared (NIR) fluorescent rhodamine derivatives, bearing two spirolactone rings and substitution of the oxygen atoms in the xanthene ring with sulfur atoms (1-S). The diastereoisomeric cis- and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service