Skip to Content
Merck
All Photos(1)

Key Documents

101893

Sigma-Aldrich

Perfluorobenzophenone

98%

Synonym(s):

Decafluorobenzophenone, Perfluorobenzophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F5COC6F5
CAS Number:
Molecular Weight:
362.12
Beilstein:
2066476
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

90-95 °C (lit.)

SMILES string

Fc1c(F)c(F)c(c(F)c1F)C(=O)c2c(F)c(F)c(F)c(F)c2F

InChI

1S/C13F10O/c14-3-1(4(15)8(19)11(22)7(3)18)13(24)2-5(16)9(20)12(23)10(21)6(2)17

InChI key

WWQLXRAKBJVNCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Perfluorobenzophenone was used in the synthesis of fluorinated acridones and acridines.

Biochem/physiol Actions

Perfluorobenzophenone alcoholic solution generates ketyl and radical anion which can be studied by electron spin resonance spectra. Perfluorobenzophenone undergoes photolysis to form ketyl (C6F5)C.OH radical.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Orthogonal Synthesis of Fluorinated Acridones and Acridines from Perfluorobenzophenone.
Del Buttero P, et al
European Journal of Organic Chemistry, 2011(12), 2265-2271 (2011)
The Photochemically Produced Ketyl of Perfluorobenzophenone: Electron Spin Resonance Studies of Solvent Effects.
Sargent FP and Bailey MG.
Canadian Journal of Chemistry, 51(19), 3211-3216 (1973)
The Electron Spin Resonance Spectra of the Photochemically Produced Perfluorobenzophenone Ketyl and Anion Radicals.
Sargent FP and Bailey MG.
Canadian Journal of Chemistry, 49(13), 2350-2352 (1971)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service