Merck
  • Gas Chromatography-Quadrupole Time-of-Flight Mass Spectrometry-Based Determination of Isotopologue and Tandem Mass Isotopomer Fractions of Primary Metabolites for (13)C-Metabolic Flux Analysis.
Gas Chromatography-Quadrupole Time-of-Flight Mass Spectrometry-Based Determination of Isotopologue and Tandem Mass Isotopomer Fractions of Primary Metabolites for (13)C-Metabolic Flux Analysis.
Analytical chemistry (2015-10-30)
Teresa Mairinger, Matthias Steiger, Justyna Nocon, Diethard Mattanovich, Gunda Koellensperger, Stephan Hann
ABSTRACT
For the first time an analytical work flow based on accurate mass gas chromatography-quadrupole time-of-flight mass spectrometry (GC-QTOFMS) with chemical ionization for analysis providing a comprehensive picture of (13)C distribution along the primary metabolism is elaborated. The method provides a powerful new toolbox for (13)C-based metabolic flux analysis, which is an emerging strategy in metabolic engineering. In this field, stable isotope tracer experiments based on, for example, (13)C are central for providing characteristic patterns of labeled metabolites, which in turn give insights into the regulation of metabolic pathway kinetics. The new method enables the analysis of isotopologue fractions of 42 free intracellular metabolites within biotechnological samples, while tandem mass isotopomer information is also accessible for a large number of analytes. Hence, the method outperforms previous approaches in terms of metabolite coverage, while also providing rich isotopomer information for a significant number of key metabolites. Moreover, the established work flow includes novel evaluation routines correcting for isotope interference of naturally distributed elements, which is crucial following derivatization of metabolites. Method validation in terms of trueness, precision, and limits of detection was performed, showing excellent analytical figures of merit with an overall maximum bias of 5.8%, very high precision for isotopologue and tandem mass isotopomer fractions representing >10% of total abundance, and absolute limits of detection in the femtomole range. The suitability of the developed method is demonstrated on a flux experiment of Pichia pastoris employing two different tracers, i.e., 1,6(13)C2-glucose and uniformly labeled (13)C-glucose.
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Sucrose, SAJ first grade
Sigma-Aldrich
Succinic acid, SAJ special grade, ≥99.5%
Sigma-Aldrich
Sucrose, JIS special grade
Sigma-Aldrich
Citric acid monohydrate, SAJ first grade, ≥99.5%
Sigma-Aldrich
Citric acid monohydrate, JIS special grade, ≥99.5%
Sigma-Aldrich
Citric acid monohydrate, ≥99.5%, suitable for amino acid analysis
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
Sigma-Aldrich
Acetic anhydride, JIS special grade
Sigma-Aldrich
D-Sedoheptulose 7-phosphate lithium salt, ≥90% (TLC)
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Dihydroxyacetone phosphate lithium salt, ≥95.0% (TLC)
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Sigma-Aldrich
DL-Malic acid, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
SAFC
L-Isoleucine
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
D-Erythrose 4-phosphate sodium salt, ≥50% (TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-2-Aminoadipic acid, ≥99%
Sigma-Aldrich
L-Homoserine
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC)