Merck
  • Persistent and emerging pollutants in the blood of German adults: Occurrence of dechloranes, polychlorinated naphthalenes, and siloxanes.
Persistent and emerging pollutants in the blood of German adults: Occurrence of dechloranes, polychlorinated naphthalenes, and siloxanes.
Environment international (2015-10-11)
Hermann Fromme, Enrique Cequier, Jun-Tae Kim, Linda Hanssen, Bettina Hilger, Cathrine Thomsen, Yoon-Seok Chang, Wolfgang Völkel
ABSTRACT
Human biomonitoring is a valid method to determine exposure, identify time trends, and monitor the effects of restrictions and measures. To characterize the recent exposure of Germans to persistent or emerging substances, we analyzed 4 dechloranes, 33 polychlorinated naphthalenes (PCNs), and 3 cyclic volatile methyl siloxanes (cVMS) in 42 plasma samples. The samples were collected from blood donors on a random selection. The median values of both Dechlorane Plus (DDC-CO) isomers were 1.23ng/gl.w. for anti- and 0.77ng/gl.w. for syn-DDC-CO. The two other dechloranes were found at lower levels. The median level of ∑-PCNs was 575pg/gl.w. (range: 101-1406pg/gl.w.). On average, the levels of PCNs in plasma were dominated by the congeners CN73, CN66/67, and CN51, which were responsible for approximately 71% of the total amount of PCNs. The cVMS octa-, deca-, and dodecamethylcyclotetrasiloxane could be determined in only some samples, with maximum values of 0.73, 0.48, and 0.79μg/l, respectively. Regarding dechloranes, our results are similar to those from other western countries but slightly lower than results from China. The levels of PCNs in German blood are similar to those observed in the U.S.A., but considerably lower than those reported for Korea. Using a preliminary TEF (toxic equivalency factor), the mean TEQ of the 9 quantifiable PCNs in Germany was low (0.36pg TEQ/gl.w.). The PCN levels in our study group are lower compared to previous studies.
Sigma-Aldrich
Hexane, suitable for HPLC
Sigma-Aldrich
Tris(trimethylsiloxy)silane, ≥98%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, for residue analysis
Sigma-Aldrich
Hexane, for residue analysis, JIS 5000
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, for residue analysis
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis