Merck
  • Assessment and modelling of Ni(II) retention by an ion-imprinted polymer: application in natural samples.
Assessment and modelling of Ni(II) retention by an ion-imprinted polymer: application in natural samples.
Journal of colloid and interface science (2015-03-17)
Véronique Lenoble, Walid Meouche, Katri Laatikainen, Cédric Garnier, Hugues Brisset, André Margaillan, Catherine Branger
ABSTRACT
Three novel Ni(II)-Ion-Imprinted Polymer (IIP) were synthesized by precipitation polymerization of ethylene glycol dimethacrylate (crosslinker) with a complex of nickel(II) and vinylbenzyl iminodiacetic acid (VbIDA). The three IIPs were prepared with various mixtures of porogen solvents: methanol, methanol/2-methoxyethanol and methanol/acetonitrile (IIP1, IIP2 and IIP3, respectively). Non-Imprinted Polymers (NIP1, NIP2 and NIP3) were prepared as control polymers in similar conditions but with pure VbIDA instead of VbIDA-Ni. These polymers were characterized by FTIR, BET, SEM and tested for their efficiency and selectivity in Ni(II) retention. The most efficient (IIP1, around 12 mg g(-1) of nickel) was then positively checked for Ni(II) retention in presence of some competing species over a wide range of concentration. Finally Ni(II) retention by IIP1 was successfully demonstrated in natural samples. The modelling of the different experiments (Langmuir, Freundlich but also PROSECE and WHAM VII, frequently used in environmental studies) allowed demonstrating the presence of completely different binding sites when considering the ion-imprinted polymer and the non-imprinted one, and therefore led to a better understanding of what the imprinting effect is.
Sigma-Aldrich
Calcium nitrate tetrahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Nitric acid, 1 M
Sigma-Aldrich
Nitric acid, SAJ first grade, 65.0-66.0%, density: 1.40
Sigma-Aldrich
Nitric acid, 0.1 M
Sigma-Aldrich
Calcium nitrate tetrahydrate, JIS special grade, ≥98.5%
Sigma-Aldrich
Iminodiacetic acid, 98%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Calcium nitrate tetrahydrate, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Hydriodic acid, 57 wt. %, distilled, 99.999% trace metals basis
Sigma-Aldrich
Vinylbenzyl chloride, purum, mixture of isomers, ≥95.0%, stabilized
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Cobalt(II) nitrate hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Cobalt(II) nitrate hexahydrate, JIS special grade, ≥98.0%
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
2-Methoxyethanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
2-Methoxyethanol, JIS special grade, ≥99.0%
Sigma-Aldrich
2-Methoxyethanol, anhydrous, 99.8%
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium nitrate, SAJ first grade, ≥99.0%