752851
Hydriodic acid
57 wt. %, distilled, 99.999% trace metals basis
Synonym(s):
Hydriotic acid
About This Item
Recommended Products
grade
distilled
Quality Level
Assay
99.999% trace metals basis
form
liquid
concentration
57 wt. %
bp
127 °C (lit.)
storage temp.
2-8°C
SMILES string
I
InChI
1S/HI/h1H
InChI key
XMBWDFGMSWQBCA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service