Merck
  • Application of quantitative NMR for purity determination of standard ACE inhibitors.
Application of quantitative NMR for purity determination of standard ACE inhibitors.
Journal of pharmaceutical and biomedical analysis (2015-06-13)
Shi Shen, Xing Yang, Yaqin Shi
ABSTRACT
This study investigated the accuracy of the quantitative NMR method for purity determination of ACE inhibitors reference standards and the discovery of two pairs of new diastereoisomers. Six types of ACE inhibitors, imidapril hydrochloride, benazepril hydrochloride, lisinopril, enalapril maleate, quinapril hydrochloride, and captopril were quantificated and validated for the qNMR method by discussing factors that affect parameters of the qNMR experiment, internal standards, integration, pH-effect, and uncertainty. The results were compared with data obtained by the mass balance method. The study found that maleic acid influenced the quantification of captopril in deuteroxide because of a chemical reaction. The mixtures of the reaction products were isolated by HPLC and structurally elucidated by NMR as two pairs of new diastereoisomers, 1-[(2S,4R)-thio-2-methylpropionyl-5-d-ethanedicarboxylicacid]-L-proline and 1-[(2S,4S)-thio-2-methylpropionyl-5-d-ethanedicarboxylicacid]-L-proline. The results showed that the accuracy and precision of quantitative (1)H NMR spectroscopy satisfied the requirements for quantitative analysis of chemical reference standards and provided a simple, rapid, and reliable method for purity determination of ACE inhibitors systematically.
Sigma-Aldrich
Maleic acid, ReagentPlus®, ≥99% (HPLC)
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, anhydrous, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, "100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, "100%", 99.96 atom % D
Sigma-Aldrich
Methanol-d4, 99 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
3,4,5-Trichloropyridine, 98%
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, NMR reference standard