Merck
  • Low-dose Bisphenol A Activates Cyp11a1 Gene Expression and Corticosterone Secretion in Adrenal Gland via the JNK Signaling Pathway.
Low-dose Bisphenol A Activates Cyp11a1 Gene Expression and Corticosterone Secretion in Adrenal Gland via the JNK Signaling Pathway.
Toxicological sciences : an official journal of the Society of Toxicology (2015-07-26)
Hsin-Chieh Lan, I-Wen Lin, Zhi-Jie Yang, Jyun-Hong Lin
ABSTRACT
Certain commonly used compounds that interfere with the functions of the endocrine system are classified as endocrine-disrupting chemicals (EDCs). Bisphenol A (BPA) is an EDC that is widely used in food containers. BPA levels in human sera are commonly observed to be approximately 1-100 nM. Compared with the effects of BPA on the gonads, its effects on the adrenal gland are poorly understood. To investigate the influence of BPA on steroidogenesis, we examined the activity of the steroidogenic gene Cyp11a1 and its regulatory pathways in mouse Y1 adrenal cortex cells. Treatment with BPA at < 100 µM did not cause cell death. However, increased promoter activity and protein expression of Cyp11a1 were induced by low doses of BPA (10-1000 nM). Moreover, BPA induced c-Jun phosphorylation, and a specific inhibitor of c-Jun N-terminal kinase (JNK) significantly suppressed BPA-induced steroidogenesis. Thus, treatment of adrenal cells with low doses of BPA activated Cyp11a1 and increased corticosterone production through the JNK/c-Jun signaling pathway. Identical results were observed in rats after BPA injection. The abnormal induction of hormone synthesis by BPA in the adrenal gland might be linked to human metabolic defects and neuropsychiatric disorders.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
8-Bromoadenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Millipore
Immobilon®-NC HATF membranes, diam. 132 mm
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Millipore
Immobilon®-FL PVDF membrane, pore size 0.45 μm
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C