Merck
  • Spiropyran-Decorated SiO₂-Pt Janus Micromotor: Preparation and Light-Induced Dynamic Self-Assembly and Disassembly.
Spiropyran-Decorated SiO₂-Pt Janus Micromotor: Preparation and Light-Induced Dynamic Self-Assembly and Disassembly.
ACS applied materials & interfaces (2015-10-22)
Qilu Zhang, Renfeng Dong, Xueyi Chang, Biye Ren, Zhen Tong
ABSTRACT
The controlled self-assembly of self-propelled Janus micromotors may give the micromotors some potential applications in many fields. In this work, we design a kind of SiO2-Pt Janus catalytic micromotor functionalized by spiropyran (SP) moieties on the surface of the SiO2 hemisphere. The spiropyran-modified SiO2-Pt Janus micromotor exhibits autonomous self-propulsion in the presence of hydrogen peroxide fuel in N,N-dimethylformamide (DMF)/H2O (1:1 in volume) mixture. We demonstrate that the self-propelled Janus micromotors can dynamically assemble into multiple motors because of the electrostatic attractions and π-π stacking between MC molecules induced by UV light irradiation (λ = 365 nm) and also quickly disassemble into mono motors when the light is switched to green light (λ = 520 nm) for the first time. Furthermore, the assembled Janus motors can move together automatically with different motion patterns propelled by the hydrogen peroxide fuels upon UV irradiation. The work provides a new approach not only to the development of the potential application of Janus motors but also to the fundamental science of reversible self-assembly and disassembly of Janus micromotors.
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
11-Bromo-1-undecanol, 98%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Piperidine, ReagentPlus®, 99%
Sigma-Aldrich
2-Hydroxy-5-nitrobenzaldehyde, 98%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Piperidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Piperidine, SAJ first grade, ≥99.0%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Piperidine, biotech. grade, ≥99.5%