Skip to Content
Merck
All Photos(1)

Key Documents

316989

Sigma-Aldrich

Hydrogen peroxide solution

contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

vapor density

1 (vs air)

Quality Level

vapor pressure

23.3 mmHg ( 30 °C)

Assay

99.999% trace metals basis

form

liquid

contains

potassium stannate as inhibitor
stabilizer

reaction suitability

reagent type: oxidant

concentration

30-32 wt. % in water, semiconductor grade

density

1.11 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hydrogen peroxide solution is a widely used oxidizing agent in a variety of applications, including the oxidation of cyanide in industrial waste samples, the removal of ammonia from semiconductor wastewater, the removal of ibuprofen from water and wastewater, and the treatment of wastewater containing five-membered heterocyclic pyrazoles[1][2][3][4]. It is one of the most widely used wet electronic chemicals[5].

Application

Hydrogen peroxide solution can be used in the substrate preparation for ultrathin polymer films[6].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MiR-34 modulates Caenorhabditis elegans lifespan via repressing the autophagy gene atg9
Yang J, et al.
Age (Dordrecht, Netherlands), 35(1), 11-22 (2013)
Integrated countercurrent reverse osmosis cascades for hydrogen peroxide ultrapurification
Abejon R, et al.
Computers & Chemical Engineering, 41, 67-76 (2012)
Comparison of hydrogen peroxide-based advanced oxidation processes for the treatment of azole-containing industrial wastewater
Li R, et al.
Chemical Engineering Journal, 425, 131785-131785 (2021)
Fabrication of large-area free-standing ultrathin polymer films
Stadermann M, et al.
Journal of Visualized Experiments, e52832-e52832 (2015)
Ibuprofen elimination from water and wastewater using sonication/ultraviolet/hydrogen peroxide/zeolite-titanate photocatalyst system
Farhadi N, et al.
Environmental Research, 198, 111260-111260 (2021)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service