Merck
  • Polyunsaturated Fat Intake Estimated by Circulating Biomarkers and Risk of Cardiovascular Disease and All-Cause Mortality in a Population-Based Cohort of 60-Year-Old Men and Women.
Polyunsaturated Fat Intake Estimated by Circulating Biomarkers and Risk of Cardiovascular Disease and All-Cause Mortality in a Population-Based Cohort of 60-Year-Old Men and Women.
Circulation (2015-06-19)
Matti Marklund, Karin Leander, Max Vikström, Federica Laguzzi, Bruna Gigante, Per Sjögren, Tommy Cederholm, Ulf de Faire, Mai-Lis Hellénius, Ulf Risérus
ABSTRACT
High intake of polyunsaturated fatty acids (PUFAs) may reduce the risk of cardiovascular disease (CVD) and mortality. Large, prospective studies including both sexes and circulating PUFAs as dietary biomarkers are needed. We investigated sex-specific associations of the major dietary PUFAs, eicosapentaenoic acid, docohexaenoic acid, linoleic acid, and α-linolenic acid, with incident CVD and all-cause mortality in a population-based cohort. PUFAs in serum cholesterol esters were measured at baseline in 60-year-old Swedish women (n=2193) and men (n=2039). With the use of national registers, 484 incident CVD events (294 men and 190 women) and 456 all-cause deaths (265 men and 191 women) were identified during follow-up (median, 14.5 years) in individuals without prior CVD at baseline. Associations of PUFAs with CVD and mortality were evaluated with Cox proportional hazard models. In multivariable-adjusted models, 1-SD increases in eicosapentaenoic acid and docohexaenoic acid were associated with lower risk of incident CVD among women (hazard ratio [HR], 0.79 [95% confidence interval (CI), 0.64-0.97] and 0.74 [95% CI, 0.61-0.89], respectively). α-Linolenic acid was associated with moderately increased CVD risk in women (HR, 1.16; 95% CI, 1.02-1.32). Inverse associations with all-cause mortality were observed for eicosapentaenoic acid and docohexaenoic acid among all participants (HR, 0.81 [95% CI, 0.72-0.91] and 0.80 [95% CI, 0.72-0.89], respectively) and for linoleic acid in men (HR, 0.73; 95% CI, 0.64-0.83). Serum linoleic acid and very-long-chain n-3 PUFAs, partly reflecting vegetable oil and fish intake, respectively, were inversely associated with all-cause mortality. Inverse associations of eicosapentaenoic acid and docohexaenoic acid with incident CVD were observed only in women.
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, for residue analysis
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, for residue analysis
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Hexane, for residue analysis, JIS 5000
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Sigma-Aldrich
Hexane, suitable for HPLC