Merck
  • Bimodal tumor-targeting from microenvironment responsive hyaluronan layer-by-layer (LbL) nanoparticles.
Bimodal tumor-targeting from microenvironment responsive hyaluronan layer-by-layer (LbL) nanoparticles.
ACS nano (2014-08-08)
Erik C Dreaden, Stephen W Morton, Kevin E Shopsowitz, Jae-Hyeok Choi, Zhou J Deng, Nam-Joon Cho, Paula T Hammond
ABSTRACT
Active targeting of nanoscale drug carriers can improve tumor-specific delivery; however, cellular heterogeneity both within and among tumor sites is a fundamental barrier to their success. Here, we describe a tumor microenvironment-responsive layer-by-layer (LbL) polymer drug carrier that actively targets tumors based on two independent mechanisms: pH-dependent cellular uptake at hypoxic tumor pH and hyaluronan-directed targeting of cell-surface CD44 receptor, a well-characterized biomarker for breast and ovarian cancer stem cells. Hypoxic pH-induced structural reorganization of hyaluronan-LbL nanoparticles was a direct result of the nature of the LbL electrostatic complex, and led to targeted cellular delivery in vitro and in vivo, with effective tumor penetration and uptake. The nanoscale drug carriers selectively bound CD44 and diminished cancer cell migration in vitro, while co-localizing with the CD44 receptor in vivo. Multimodal targeting of LbL nanoparticles is a powerful strategy for tumor-specific cancer diagnostics and therapy that can be accomplished using a single bilayer of polyamine and hyaluronan that, when assembled, produce a dynamic and responsive cell-particle interface.
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
DAPI, for nucleic acid staining