Merck
  • Synthesis of hollow mesoporous silica nanoparticles with tunable shell thickness and pore size using amphiphilic block copolymers as core templates.
Synthesis of hollow mesoporous silica nanoparticles with tunable shell thickness and pore size using amphiphilic block copolymers as core templates.
Dalton transactions (Cambridge, England : 2003) (2014-06-25)
Xiaojun Zhou, Xiao Cheng, Wei Feng, Kexin Qiu, Liang Chen, Wei Nie, Zhiqi Yin, Xiumei Mo, Hongsheng Wang, Chuanglong He
ABSTRACT
This paper presents a facile method for the fabrication of uniform hollow mesoporous silica nanoparticles (HMSNs) with tunable shell thickness and pore size. In this method, a series of amphiphilic block copolymers of polystyrene-b-poly (acrylic acid) (PS-b-PAA) with different hydrophobic block (PS) lengths were first synthesized via atom transfer radical polymerization (ATRP). The as-synthesized PS-b-PAA and cetyltrimethylammonium bromide (CTAB) were subsequently used as co-templates to fabricate HMSNs. This approach allows the control of shell thickness and pore size distribution of the synthesized HMSNs simply by changing the amounts of PS-b-PAA and CTAB, respectively. In vitro cytotoxicity and hemolysis assays demonstrated that the synthesized HMSNs had a low and shell thickness-dependent cytotoxicity and hemolytic activity. Therefore, these HMSNs have great potential for biomedical applications due to their good biocompatibility and ease of synthesis.
Sigma-Aldrich
Sulfanilic acid, JIS special grade, 99.0-100.5%
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer
Sigma-Aldrich
Styrene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Supelco
Styrene, analytical standard
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Supelco
Styrene, analytical standard
Sigma-Aldrich
Sulfanilic acid, puriss. p.a., ≥99.0% (T)
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Sulfanilic acid, ACS reagent, 99%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
USP
Sulfanilic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Supelco
Sulfanilic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonia solution, 7 N in methanol