Merck
  • Ephrin/Ephrin receptor expression in ammonia-treated rat astrocytes and in human cerebral cortex in hepatic encephalopathy.
Ephrin/Ephrin receptor expression in ammonia-treated rat astrocytes and in human cerebral cortex in hepatic encephalopathy.
Neurochemical research (2014-07-30)
Karmela Sobczyk, Markus S Jördens, Ayse Karababa, Boris Görg, Dieter Häussinger
ABSTRACT
Hepatic encephalopathy (HE) represents a neuropsychiatric syndrome, which evolves as a consequence of a low grade cerebral edema and a concomitant oxidative/nitrosative stress response. Ephrin receptors (EphR) and their ligands (ephrins) regulate astrocytic glutamate uptake and gliotransmitter release thereby governing neurotransmission, but their role in HE and ammonia toxicity is unclear. We therefore tested effects of ammonia on expression levels of EphR/ephrin isoforms in cultured rat astrocytes and analysed underlying mechanisms. NH4Cl induced mRNA expression changes of several EphR/ephrin isoforms in a methionine sulfoximine-, NADPH oxidase- and NO synthase-dependent manner in cultured astrocytes. A prominent upregulation was noted for EphR A4 mRNA and protein in NH4Cl-treated astrocytes. NH4Cl-treatment decreased EphR A4 molecular mass to similar extent as found in astrocytes treated with the N-glycosylation inhibitor tunicamycin. Knockdown of EphR A4 by siRNA, or treating astrocytes with NH4Cl or tunicamycin abolished fibroblast growth factor-induced and EphR A4-dependent astrocyte proliferation. NH4Cl-treatment also decreased GLAST mRNA levels in cultured astrocytes. This effect was sensitive to inhibitors of NAPDH oxidase or glutamine synthetase, but was insensitive to siRNA-mediated EphR A4 knockdown. Eph/ephrin gene expression changes were also found in post mortem brain samples of cirrhotic patients without or with HE compared to controls suggesting a potential in vivo relevance of the present findings. The present study suggests that ammonia modulates EphR/ephrin signaling in astrocytes and in the brain of cirrhotic patients with HE with potential implications for deranged neurotransmission in HE.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Supelco
L-Methionine sulfoximine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Methionine sulfoximine, suitable for cell culture
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Supelco
Ammonium ion solution for ISE, for ion-selective electrodes, 1000 mg/kg N, analytical standard (for ion-selective electrodes)