Merck
  • GliA in Aspergillus fumigatus is required for its tolerance to gliotoxin and affects the amount of extracellular and intracellular gliotoxin.
GliA in Aspergillus fumigatus is required for its tolerance to gliotoxin and affects the amount of extracellular and intracellular gliotoxin.
Medical mycology (2014-05-23)
Dan-Ni Wang, Takahito Toyotome, Yasunori Muraosa, Akira Watanabe, Tuya Wuren, Somnuk Bunsupa, Kaori Aoyagi, Mami Yamazaki, Masahiko Takino, Katsuhiko Kamei
ABSTRACT
Gliotoxin is an important virulence factor of Aspergillus fumigatus. Although GliA putatively belongs to the major facilitator superfamily in the gliotoxin biosynthesis cluster, its roles remain unclear. To determine the function of GliA, we disrupted gliA in A. fumigatus. gliA disruption increased the susceptibility of A. fumigatus to gliotoxin. The gliT and gliA double-disrupted mutant had even higher susceptibility to gliotoxin than each individual disruptant. The extracellular release of gliotoxin was greatly decreased in the gliA disruptant. Mice infected with the gliA disruptant of A. fumigatus showed higher survival rates than those infected with the parent strain. These results strongly indicate that GliA, in addition to GliT, plays a significant role in the tolerance to gliotoxin and protection from extracellular gliotoxin in A. fumigatus by exporting the toxin. This also allows the fungus to evade the harmful effect of its own gliotoxin production. Moreover, GliA contributes to the virulence of A. fumigatus through gliotoxin secretion.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Hydrocortisone acetate, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone acetate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone 21-acetate, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone 21-acetate, ≥98% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent