Skip to Content
Merck
All Photos(4)

Key Documents

13433

Sigma-Aldrich

Sodium bicarbonate

puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder

Synonym(s):

Sodium hydrogen carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaHCO3
CAS Number:
Molecular Weight:
84.01
Beilstein:
4153970
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38020213
PubChem Substance ID:
NACRES:
NA.21
Assay:
99.0-100.5%
form:
powder
Pricing and availability is not currently available.

grade

puriss.

Quality Level

Assay

99.0-100.5%

form

powder

quality

meets analytical specification of Ph. Eur., BP, USP, FCC, E500

impurities

ammonia (NH3), in accordance
organic volatile impurities, complies
residual solvents, complies
water-insoluble matter, complies
≤0.002% ammonium (NH4), complies

loss

≤0.2% loss on drying (silica gel)

pH

≤8.6 (20 °C, 5%)

pKa (25 °C)

(1) 6.37, (2) 10.25 (carbonic acid)

density

2.16 g/mL at 25 °C (lit.)

anion traces

carbonate (CO32-): in accordance
chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤0.5 mg/kg
As: ≤2 mg/kg
Ca: ≤100 mg/kg
Fe: ≤10 mg/kg
Hg: ≤1 mg/kg
K: ≤100 mg/kg
Pb: ≤2 mg/kg

suitability

complies for appearance of solution

SMILES string

[Na+].OC([O-])=O

InChI

1S/CH2O3.Na/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

UIIMBOGNXHQVGW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium bicarbonate may be involved in the fabrication of silica based monolith columns which can be used in capillary electrochromatography.[1] It can also be used to maintain the pH of the column for the separation of proteins via ion exchange displacement chromatography.[2]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silica-based monoliths for capillary electrochromatography: Methods of fabrication and their applications in analytical separations
Allen D and Rassi EZ
Electrophoresis, 24, 3962-3976 (2003)
Use of the sodium salt of poly (vinylsulfonic acid) as a low-molecular-weight displacer for protein separations by ion-exchange displacement chromatography
Jen D.C.S and Pinto.GN
Journal of Chromatography A, 519(1), 87-98 (1990)
Shay P McGuinness et al.
Critical care medicine, 41(7), 1599-1607 (2013-05-21)
Cardiac surgery-associated acute kidney injury occurs in up to 50% of patients and is associated with increased mortality and morbidity. This study aimed to discover if perioperative urinary alkalinization with sodium bicarbonate infusion reduces the prevalence of cardiac surgery-associated acute
Joshua C Tremblay et al.
The Journal of physiology, 593(3), 723-737 (2014-11-25)
Blood flow through intrapulmonary arteriovenous anastomoses (IPAVA) is increased by acute hypoxia during rest by unknown mechanisms. Oral administration of acetazolamide blunts the pulmonary vascular pressure response to acute hypoxia, thus permitting the observation of IPAVA blood flow with minimal
Sophia Zoungas et al.
Annals of internal medicine, 151(9), 631-638 (2009-11-04)
Intravenous sodium bicarbonate has been proposed to reduce the risk for contrast-induced nephropathy (CIN). To determine the effect of sodium bicarbonate on the risk for CIN. MEDLINE, PubMed, EMBASE, and the Cochrane Central Register of Controlled Trials from 1950 to

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service