Merck
  • An LC-MS/MS method for the simultaneous determination of goserelin and testosterone in rat plasma for pharmacokinetic and pharmacodynamic studies.
An LC-MS/MS method for the simultaneous determination of goserelin and testosterone in rat plasma for pharmacokinetic and pharmacodynamic studies.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-07-20)
Shu Zhang, Jiangbin Han, Guangyi Leng, Xin Di, Chunjie Sha, Xuemei Zhang, Youxin Li, Wanhui Liu
ABSTRACT
A liquid chromatography-electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) method was developed, using testosterone-d3 as a surrogate analyte, for the simultaneous quantification of goserelin and testosterone in rat plasma. According to this method, the pharmacokinetic and pharmacodynamic data were obtained from a single plasma sample aliquot. The method involved the addition of alarelin as an internal standard (IS) for goserelin and testosterone-(13)C3 for testosterone or testosterone-d3. The conditions for the separation of these two compounds were achieved on a ZORBAX Eclipse Plus C18 column (Agilent, 2.1 × 50 mm, 1.8 μm, Stockport, UK) in a single chromatographic run at a flow rate of 400 μL/min. In order to minimize interferences of complex matrix, the extraction of plasma consisted of a protein precipitation step using methanol, followed by purification using an Oasis(®) HLB solid-phase extraction column. The method was validated in the concentration range of 0.01-30.0 ng/mL for goserelin and 0.05-30.0 ng/mL for testosterone-d3, respectively. The within- and between-run precisions were 1.7-9.2% and 2.1-6.9%, respectively. The within- and between-run accuracies were -1.8 to 5.3% and -4.9 to 4.0%, respectively. This accurate and highly specific assay provides a useful method to evaluate the pharmacokinetics and pharmacodynamics of goserelin in rats.
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Testosterone, European Pharmacopoeia (EP) Reference Standard
Supelco
Testosterone-2,3,4-13C3 solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Testosterone-2,3,4-13C3 solution, 0.1 mg/mL in methanol, 99 atom % 13C, 98% (CP)
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Testosterone, ≥98%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)