Merck
  • Cofilin-phosphatase slingshot-1L (SSH1L) is over-expressed in pancreatic cancer (PC) and contributes to tumor cell migration.
Cofilin-phosphatase slingshot-1L (SSH1L) is over-expressed in pancreatic cancer (PC) and contributes to tumor cell migration.
Cancer letters (2015-02-17)
Yufeng Wang, Yasuhiro Kuramitsu, Takao Kitagawa, Byron Baron, Shigefumi Yoshino, Shin-Ichiro Maehara, Yoshihiko Maehara, Masaaki Oka, Kazuyuki Nakamura
ABSTRACT
Slingshot-1L (SSH1L), a cofilin-phosphatase, plays a role in actin dynamics and cell migration by reactivating cofilin-1. However, the expression of SSH1L in malignant diseases is poorly understood. The overexpression of SSH1L in cancerous tissue compared to the matched surrounding non-cancerous tissues from patients with late stages (III-IV) of PC was detected in 90% (9/10) of cases by western blotting. The expression of SSH1L was shown to be upregulated in tumor cells from 10.7% (11/102) of patients with pancreatic cancer (PC) by immunohistochemistry (IHC). The positive rate of SSH1L in patients with PC at stage VI (TNM) categorized as grade 3 was of 50% (2/4) and 15% (6/40), respectively. Moreover, SSH1L expression was shown to be up-regulated in the PC cell lines (KLM1, PANC-1 and MIAPaCa-2) with high metastatic potential. Loss of SSH1L expression was associated with an increase in the phosphorylation of cofilin-1 at serine-3 and further inhibited cell migration (but not proliferation) in KLM1, PANC-1 and MIAPaCa-2. Actin polymerization inhibitor cytochalasin-D was sufficient to abrogate cell migration of PC without changing SSH1L expression. These results reveal that SSH1L is upregulated in a subset of PCs and that the SSH1L/cofilin-1 signal pathway is associated positively in PC with cell migration. Our study may thus provide potential targets to prevent and/or treat PC invasion and metastasis in patients with SSH1L-positive PC.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium fluoride, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder