Merck
  • Vine-shoot waste aqueous extracts for re-use in agriculture obtained by different extraction techniques: phenolic, volatile, and mineral compounds.
Vine-shoot waste aqueous extracts for re-use in agriculture obtained by different extraction techniques: phenolic, volatile, and mineral compounds.
Journal of agricultural and food chemistry (2014-10-23)
Rosario Sánchez-Gómez, Amaya Zalacain, Gonzalo L Alonso, M Rosario Salinas
ABSTRACT
Vine-shoots are an important waste in all viticulture areas that should be re-used with innovative applications. The aim of this work was to produce Airén waste vine-shoot aqueous extracts by four solid-liquid extraction techniques such as conventional solid-liquid extraction (CSLE), solid-liquid dynamic extraction (SLDE-Naviglio), microwave extraction (ME), and pressurized solvent extraction (PSE). Their chemical composition was studied in terms of phenolic, volatile, and mineral compounds. The highest concentrated extracts corresponded to CSLE and SLDE-Naviglio, independent of the conditions tested. The CSLE extracts had the highest flavanols, phenolic acids, and stilbenes contents. The volatile composition, quantified for first time in this work, shows that furanic compounds were the most abundant. All extracts showed an interesting mineral content, which may be assimilated by plants. These results show the agricultural potential of Airén vine-shoot waste aqueous extracts to be used as grape biostimulants and/or foliar fertilizer.
Sigma-Aldrich
Benzaldehyde, SAJ special grade, ≥98.0%
Sigma-Aldrich
Guaiacol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Furfural, SAJ first grade, ≥99.0%
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Linalool, analytical standard
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
Decanoic acid, ≥98.0%
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Decanoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Hexanoic acid, purum, ≥98.0% (GC)
Supelco
Decanoic acid, analytical standard
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
5-Methylfurfural, ReagentPlus®, 99%
Sigma-Aldrich
Linalool, 97%
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Supelco
Benzaldehyde, analytical standard
Sigma-Aldrich
Furfural, 99%
Sigma-Aldrich
Furfural, natural, ≥98%, FCC, FG
Sigma-Aldrich
Linalyl acetate, ≥97%, FCC, FG
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
Decanoic acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Furfural, ACS reagent, 99%
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
Furfural, ≥98%, FCC, FG