Skip to Content
Merck
All Photos(2)

Key Documents

G5502

Sigma-Aldrich

Guaiacol

oxidation indicator

Synonym(s):

2-Methoxyphenol, Catechol monomethyl ether, Pyrocatechol monomethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)C6H4OH
CAS Number:
Molecular Weight:
124.14
Beilstein:
508112
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

vapor density

4.27 (vs air)

vapor pressure

0.11 mmHg ( 25 °C)

Assay

≥98.0%

form

liquid
solid

refractive index

n20/D 1.543 (lit.)

bp

205 °C (lit.)

mp

26-29 °C (lit.)

density

1.129 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1O

InChI

1S/C7H8O2/c1-9-7-5-3-2-4-6(7)8/h2-5,8H,1H3

InChI key

LHGVFZTZFXWLCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Guaiacol, a phenolic compound, acts as a scavenger of superoxide radicals. It is produced from guaiacum or wood creosote. Guaiacol acts as a precursor to different favoring agents like, vanillin and roasted coffee. It is used as an antiseptic, analgesic and sedative drug for the treatment of dental pulp and periodontal tissue. Guaiacol enhances cell proliferation in vivo.

Application

Guaiacol has been used:
  • as a substrate for peroxidase enzyme to determine its activity
  • as a substrate to determine laccase activity
  • as a reagent in peroxidase (POD) activity test

Biochem/physiol Actions

Guaiacol has an ability to inhibit prostaglandin biosynthesis, without causing damage to the gastrointestinal system, unlike classic NSAID (nonsteroidal anti-inflammatory drugs).
Guaiacol, along with 2,4,6-trichloroanisole, is responsible for cork taint in wine. A method has been developed for extraction and quantitation of the two compounds.

Caution

May be liquid at room temperature.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Increased Levels of Antinutritional and/or Defense Proteins Reduced the Protein Quality of a Disease-Resistant Soybean Cultivar
Sousa DOB, et al.
Nutrients, 7(7), 6038-6054 (2015)
Constituent profiles
Essential Oil Safety-A Guide for Health Care Professionals (2014)
Measurements of Proline and Malondialdehyde Content and Antioxidant Enzyme Activities in Leaves of Drought Stressed Cotton
Chen T and Zhang B
Plant & Cell Physiology (2015)
Kinetic analysis and structural studies of a high-efficiency laccase from Cerrena sp. RSD 1
Wu MH, et al.
FEBS Open Bio, 8(8), 1230-1246 (2018)
Antimicrobial Activity of Newly Prepared Endodontic Biosealer on Enterococcus Faecalis-An in vitro study
AL-Jubori SH, et al.
International journal of enhanced research in science technology and engineering. (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service