Merck
  • Ultra-high performance liquid chromatography-quadrupole time-of-flight mass spectrometry to identify contaminants in water: an insight on environmental forensics.
Ultra-high performance liquid chromatography-quadrupole time-of-flight mass spectrometry to identify contaminants in water: an insight on environmental forensics.
Journal of chromatography. A (2014-05-06)
Ana Masiá, Julian Campo, Cristina Blasco, Yolanda Picó
ABSTRACT
Ultra-high pressure liquid chromatography-quadrupole time-of-flight mass spectrometry (UHPLC-QqTOF-MS) acquiring full scan MS data for quantification, and automatic data dependent information product ion spectra (IDA-MS/MS) without any predefinition of the ions by the user was checked for identifying organic contaminants in water samples. The use of a database with more than 2000 compounds achieved high confidence results for a wide number of contaminants based upon retention time, accurate mass, isotopic pattern and MS/MS library searching. More than 20 contaminants, mostly pharmaceuticals, but also mycotoxins and polyphenols were unambiguously identified. Furthermore, the combination of statistical data analysis using principal component analysis (PCA) followed by empirical formula calculation, on-line database searching and MS/MS fragment ion interpretation achieves not only the successful detection of unknown contaminants but also the selection of those relevant to different types of waters. Unknown compounds, such as C₂₀H₃₄O₃, were identified in waste water showing the prospects of this technique. A group of 42 currently used pesticides were selected as target compounds to evaluate the quantitative possibilities. Mean recoveries and percentage relative standard deviation (RSD) were 48-79% (4-20% RSD). The limit of detections ranged from 0.02 to 2 ng L(-1), with a validated limit of quantification of 2 ng L(-1) for water after solid-phase (SPE) isolation and concentration. The quantitative data obtained using UHPLC-QqTOF-MS were compared with those obtained using conventional LC-MS/MS with a triple quadrupole (QqQ).
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Ammonium formate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Malathion, certified reference material, TraceCERT®
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Malathion, United States Pharmacopeia (USP) Reference Standard
Malathion, European Pharmacopoeia (EP) Reference Standard
Supelco
Isoproturon, PESTANAL®, analytical standard
Supelco
Propazine, PESTANAL®, analytical standard
Supelco
Ethion, PESTANAL®, analytical standard
Supelco
Methanol, analytical standard
Supelco
Dimethoate, PESTANAL®, analytical standard
Supelco
Malathion, PESTANAL®, analytical standard
Supelco
Propanil, PESTANAL®, analytical standard
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Parathion, PESTANAL®, analytical standard
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ammonium formate, reagent grade, 97%