Merck
  • Development of 4-hexadecyl-4,7-diaza-1,10-decanedithiol (HDD) kit for the preparation of the liver cancer therapeutic agent Re-188-HDD/lipiodol.
Development of 4-hexadecyl-4,7-diaza-1,10-decanedithiol (HDD) kit for the preparation of the liver cancer therapeutic agent Re-188-HDD/lipiodol.
Nuclear medicine and biology (2014-12-30)
Vinay Kumar Banka, Sung-Hyun Moon, Jae Min Jeong, Sudhakara Reddy Seelam, Yun-Sang Lee, Young Joo Kim, Dong Soo Lee, June-Key Chung
ABSTRACT
A lipiodol solution of (188)Re-4-hexadecyl-2,2,9,9-tetramethyl-4,7-diaza-1,10-decanedithiol (HTDD) has been successfully developed for liver cancer therapy; however, its preparation requires a multi-step synthesis and it is characterized by a low labeling yield. We synthesized a new compound, 4-hexadecyl-4,7-diaza-1,10-decanedithioacetate (AHDD), without gem dimethyl groups to address these issues. AHDD was formulated into a kit and was labeled with (188)Re. Biodistribution study was performed using normal BALB/c mice. The kit was labeled with (188)Re with a high efficiency (98.8±0.2%). After extraction with lipiodol, the overall yield of (188)Re-HDD/lipiodol was as high as 90.2±2.6%. A comparative biodistribution study of (188)Re-HTDD and (188)Re-HDD was performed in normal mice after intravenous injection. The lungs were identified as the main uptake site due to capillary-blockage. (188)Re-HDD/lipiodol showed a significantly higher lung uptake than that of (188)Re-HTDD/lipiodol (p<0.05). The newly synthesized (188)Re-HDD/lipiodol showed improved radiolabeling yield and biodistribution results compared to (188)Re-HTDD/lipiodol, and may therefore be more suitable for liver cancer therapy.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
1-Iodohexadecane, contains copper as stabilizer, 95%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard