Merck
  • Cell membrane integrity in myotonic dystrophy type 1: implications for therapy.
Cell membrane integrity in myotonic dystrophy type 1: implications for therapy.
PloS one (2015-03-24)
Anchel González-Barriga, Julia Kranzen, Huib J E Croes, Suzanne Bijl, Walther J A A van den Broek, Ingeborg D G van Kessel, Baziel G M van Engelen, Judith C T van Deutekom, Bé Wieringa, Susan A M Mulders, Derick G Wansink
ABSTRACT
Myotonic Dystrophy type 1 (DM1) is a multisystemic disease caused by toxic RNA from a DMPK gene carrying an expanded (CTG•CAG)n repeat. Promising strategies for treatment of DM1 patients are currently being tested. These include antisense oligonucleotides and drugs for elimination of expanded RNA or prevention of aberrant binding to RNP proteins. A significant hurdle for preclinical development along these lines is efficient systemic delivery of compounds across endothelial and target cell membranes. It has been reported that DM1 patients show elevated levels of markers of muscle damage or loss of sarcolemmal integrity in their serum and that splicing of dystrophin, an essential protein for muscle membrane structure, is abnormal. Therefore, we studied cell membrane integrity in DM1 mouse models commonly used for preclinical testing. We found that membranes in skeletal muscle, heart and brain were impermeable to Evans Blue Dye. Creatine kinase levels in serum were similar to those in wild type mice and expression of dystrophin protein was unaffected. Also in patient muscle biopsies cell surface expression of dystrophin was normal and calcium-positive fibers, indicating elevated intracellular calcium levels, were only rarely seen. Combined, our findings indicate that cells in DM1 tissues do not display compromised membrane integrity. Hence, the cell membrane is a barrier that must be overcome in future work towards effective drug delivery in DM1 therapy.
Product Description
Butyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Supelco
Butylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Butyl 4-hydroxybenzoate, SAJ first grade, ≥99.0%
USP
Butylparaben, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99.0% (GC)