Merck
FADD is essential for glucose uptake and survival of thymocytes.
Biochemical and biophysical research communications (2014-08-01)
Xiang-Yu Zhang, Bing-Ya Yang, Jia-Yu Wang, Xuan Mo, Jing Zhang, Zi-Chun Hua
ABSTRACT
Fas-associated protein with death domain (FADD) has been implicated in T lymphocytes, but the nature of FADD-dependent mechanism in early T cell development has not been completely elucidated. In this study, using T-cell specific deletion mice, we observed that FADD deficiency in thymocytes led to increased apoptosis and reduced cell numbers, which may be attributed to the reduction of Glut1 expression and correspondingly decreased glucose uptake. Furthermore, an abnormal transduction of Akt signaling was discovered in FADD(-/-) thymocytes, which may be responsible for the declined Glut1 expression. Collectively, our results demonstrate the new function of FADD in glucose metabolism and survival of early T cells.
Product Description
Butyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Butyl 4-hydroxybenzoate, SAJ first grade, ≥99.0%
Supelco
Butylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, ≥97% (HPLC)
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99.0% (GC)
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
USP
Butylparaben, United States Pharmacopeia (USP) Reference Standard