Merck
  • Neutrophil migration towards C5a and CXCL8 is prevented by non-steroidal anti-inflammatory drugs via inhibition of different pathways.
Neutrophil migration towards C5a and CXCL8 is prevented by non-steroidal anti-inflammatory drugs via inhibition of different pathways.
British journal of pharmacology (2014-03-07)
Maria Bertolotto, Paola Contini, Luciano Ottonello, Aldo Pende, Franco Dallegri, Fabrizio Montecucco
ABSTRACT
Non-steroidal anti-inflammatory drugs (NSAIDs) have been shown to induce PG-independent anti-inflammatory actions. Here, we investigated the role of three different NSAIDs (naproxen, ibuprofen and oxaprozin) on neutrophil responses to CXCL8 and C5a. Human neutrophils were isolated from healthy volunteers by dextran and Ficoll-Hypaque density gradients. Neutrophils were pre-incubated with different concentrations (1-100 µM) of NSAIDs or kinase inhibitors. Neutrophil degranulation into supernatants was tested by elisa and zymography. Neutrophil chemotaxis was determined using Boyden chambers. F-actin polymerization was determined by Alexa-Fluor 488-conjugated phalloidin fluorescent assay. Integrin expression was assessed by flow cytometry. The phosphorylation of intracellular kinases was studied by Western blot. Pretreatment with NSAIDs did not affect neutrophil degranulation, but inhibited neutrophil migration and polymerization of F-actin, in response to CXCL8 and C5a. Pretreatment with different NSAIDs prevented C5a-induced integrin (CD11b) up-regulation, while only ibuprofen reduced CXCL8-induced CD11b up-regulation. Pre-incubation with naproxen or oxaprozin, but not ibuprofen, inhibited the PI3K/Akt-dependent chemotactic pathways. Both endogenous (released in cell supernatants) or exogenous (added to cell cultures) PGE2 did not affect C5a- or CXCL8-induced activities. Short-term incubation with NSAIDs did not affect neutrophil PGE2 release. Treatment with NSAIDs reduced C5a- and CXCL8-induced neutrophil migration and F-actin polymerization via different mechanisms. Inhibition by ibuprofen was associated with integrin down-regulation, while naproxen and oxaprozin blocked the PI3K/Akt pathway. Both NSAID actions were independent of COX inhibition and PGE2 release.
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ibuprofen
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Human MMP-8 ELISA Kit, for serum, plasma, conditioned medium, urine
Sigma-Aldrich
Rat MMP-8 ELISA Kit, for serum, plasma, cell culture supernatant
Sigma-Aldrich
Interleukin-8 human, ≥98% (SDS-PAGE and HPLC), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Matrix Metalloproteinase-9 human, recombinant, >90% (SDS-PAGE), buffered aqueous solution
Dinoprostone, European Pharmacopoeia (EP) Reference Standard