Merck
  • Biological effects of sol-gel derived ZrO2 and SiO2/ZrO2 coatings on stainless steel surface--In vitro model using mesenchymal stem cells.
Biological effects of sol-gel derived ZrO2 and SiO2/ZrO2 coatings on stainless steel surface--In vitro model using mesenchymal stem cells.
Journal of biomaterials applications (2014-07-31)
Agnieszka Smieszek, Anna Donesz-Sikorska, Jakub Grzesiak, Justyna Krzak, Krzysztof Marycz
ABSTRACT
The objective of this study was to determine biocompatibility of zirconia-based coatings obtained by the sol-gel method. Two matrices, ZrO2 and SiO2/ZrO2, were created and applied on stainless steel type 316L with dip-coating technique. The morphology and topography of biomaterials' surface were characterized using energy-dispersive X-ray spectroscopy and atomic force microscopy, while chemical composition was analyzed by Raman spectroscopy. Additionally, wettability and surface free energy were characterized. Biocompatibility of obtained biomaterials was evaluated using an in vitro model employing mesenchymal stem cells (MSCs) of adipose and bone marrow origin. Biological analysis included determination of proliferation activity and morphology of MSCs in cultures on synthesized biomaterials. Osteoinductive properties of biomaterials were determined both in non-osteogenic, as well as osteogenic conditions. The results showed that investigated biomaterials exerted different impact on MSCs. Biomaterial with ZrO2 layer was more biocompatible for adipose-derived MSCs, while SiO2/ZrO2 layer promoted proliferation of bone marrow derived MSCs. Moreover, hybrid coating exhibited greater osteoinductive properties than ZrO2 coating, both on cultures with adipose-derived stromal (stem) cells and bone marrow stromal cells. Observed biological effects may result not only from different chemical composition, but also from diverse wettability. The ZrO2 coating was characterized as hydrophobic layer, while SiO2/ZrO2 exhibited hydrophilic properties. The results obtained suggest that behavior of MSCs in response to the biomaterial may vary depending on their origin, therefore we postulate, that screening analysis of implants' biocompatibility, should incorporate model applying both adipose- and bone marrow derived MSCs.
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
Diiodomethane, ReagentPlus®, 99%, contains copper as stabilizer
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
Diiodomethane, SAJ first grade, ≥98.0%
Sigma-Aldrich
Tetraethyl orthosilicate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Human Osteopontin ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Mouse Osteopontin ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fluorescein, for fluorescence, free acid