Merck
  • Poly-3-hydroxybutyrate-co-3-hydroxyvalerate containing scaffolds and their integration with osteoblasts as a model for bone tissue engineering.
Poly-3-hydroxybutyrate-co-3-hydroxyvalerate containing scaffolds and their integration with osteoblasts as a model for bone tissue engineering.
Journal of biomaterials applications (2015-01-17)
Sai Zhang, Molamma P Prabhakaran, Xiaohong Qin, Seeram Ramakrishna
ABSTRACT
Nano/micro engineered polymeric materials offer expansive scope of biomimetic scaffolds for bone tissue engineering especially those involving electrospun biodegradable nanofibers incorporated with inorganic nanoparticles, thus mimicking the extracellular matrix of bone both structurally and chemically. For the first time, poly-3-hydroxybutyrate-co-3-hydroxyvalerate containing natural poly-(α, β)-DL-aspartic acid and inorganic hydroxyapatite nanofibers were fabricated using poly-3-hydroxybutyrate-co-3-hydroxyvalerate: poly-(α, β)-DL-aspartic acid at a ratio of 80:20 (w/w) added with 1% (w/v) of hydroxyapatite, by the process of electrospinning. The surface morphology, chemical, and mechanical properties of electrospun poly-3-hydroxybutyrate-co-3-hydroxyvalerate, poly-3-hydroxybutyrate-co-3-hydroxyvalerate/poly-(α, β)-DL-aspartic acid, and poly-3-hydroxybutyrate-co-3-hydroxyvalerate/poly-(α, β)-DL-aspartic acid/hydroxyapatite nanofibers were characterized by using field emission scanning electron microscope, Fourier transform infrared spectroscopy, and tensile tester, respectively. Human fetal osteoblasts were cultured on different nanofibrous scaffolds for evaluating the cell proliferation, alkaline phosphatase activity, and mineralization. Cells on poly-3-hydroxybutyrate-co-3-hydroxyvalerate/poly-(α, β)-DL-aspartic acid/hydroxyapatite scaffolds demonstrated higher proliferation (30.10%) and mineral deposition (37.60%) than the cells grown on pure poly-3-hydroxybutyrate-co-3-hydroxyvalerate scaffolds. Obtained results highlight the synergistic effect of poly-3-hydroxybutyrate-co-3-hydroxyvalerate, poly-(α, β)-DL-aspartic acid, and hydroxyapatite towards the enhancement of the osteoinductivity and osteoconductivity of human fetal osteoblasts, demonstrating the appropriate physicochemical and biological properties of poly-3-hydroxybutyrate-co-3-hydroxyvalerate/poly-(α, β)-DL-aspartic acid/hydroxyapatite nanofibers to function as a substrate for bone tissue regeneration.
Sigma-Aldrich
Carbon nanofibers, graphitized, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Carbon nanofibers, pyrolitically stripped, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Phenylacetic acid, 99%
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Methane-12C, 13C-depleted, 99.9 atom % 12C
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Supelco
N,O-Bis(trimethylsilyl)acetamide, for GC derivatization, LiChropur, ≥98.5% (GC)
Supelco
Hexamethyldisilazane, for GC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Hexamethyldisilazane, Wacker Chemie AG, produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, foil, 50x50mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm