Merck
  • (125)I-Labelled 2-Iodoestrone-3-sulfate: synthesis, characterization and OATP mediated transport studies in hormone dependent and independent breast cancer cells.
(125)I-Labelled 2-Iodoestrone-3-sulfate: synthesis, characterization and OATP mediated transport studies in hormone dependent and independent breast cancer cells.
Nuclear medicine and biology (2014-12-30)
Nilasha Banerjee, T Robert Wu, Jason Chio, Ryan Kelly, Karin A Stephenson, John Forbes, Christine Allen, John F Valliant, Reina Bendayan
ABSTRACT
Organic Anion Transporting Polypeptides (OATP) are a family of membrane associated transporters that facilitate estrone-3-sulphate (E3S) uptake by hormone dependent, post-menopausal breast cancers. We have established E3S as a potential ligand for targeting hormone dependent breast cancer cells, and in this study sought to prepare and investigate radioiodinated E3S as a tool to study the OATP system. 2- and 4-Iodoestrone-3-sulfates were prepared from estrone via aromatic iodination followed by a rapid and high yielding sulfation procedure. The resulting isomers were separated by preparative HPLC and verified by (1)H NMR and analytical HPLC. Transport studies of 2- and 4-[(125)I]-E3S were conducted in hormone dependent (i.e. MCF-7) and hormone independent (i.e. MDA-MB-231) breast cancer cells in the presence or absence of the specific transport inhibitor, bromosulfophthalein (BSP). Cellular localization of OATP1A2, OATP2B1, OATP3A1 and OATP4A1 were determined by immunofluorescence analysis using anti-Na(+)/K(+) ATPase-α (1:100 dilution) and DAPI as plasma membrane and nuclear markers, respectively. Significantly (p<0.01) higher total accumulation of 2-[(125)I]-E3S was observed in hormone dependent MCF-7 as compared to hormone independent MDA-MB-231 breast cancer cells. In contrast 4-[(125)I]-E3S did not show cellular accumulation in either case. The efficiency of 2-[(125)I]-E3S transport (expressed as a ratio of Vmax/Km) was 2.4 times greater in the MCF-7 as compared to the MDA-MB-231 breast cancer cells. OATP1A2, OATP3A1 and OATP4A1 expression was localized in plasma membranes of MCF-7 and MDA-MB-231 cells confirming the functional role of these transporters in radioiodinated E3S cellular uptake. An efficient method for the preparation of 2- and 4-[(125)I]-E3S was developed and where the former demonstrated potential as an in vitro probe for the OATP system. The new E3S probe can be used to study the OATP system and as a platform to create radiopharmaceuticals for imaging breast cancer.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Cerium(III) sulfate, ≥99.99% trace metals basis
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Trifluoroacetic acid, SAJ special grade, ≥99.0%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)