Merck
  • Protein kinase-D1 overexpression prevents lipid-induced cardiac insulin resistance.
Protein kinase-D1 overexpression prevents lipid-induced cardiac insulin resistance.
Journal of molecular and cellular cardiology (2014-09-01)
Ellen Dirkx, Guillaume J J M van Eys, Robert W Schwenk, Laura K M Steinbusch, Nicole Hoebers, Will A Coumans, Tim Peters, Ben J Janssen, Boudewijn Brans, Andreas T Vogg, Dietbert Neumann, Jan F C Glatz, Joost J F P Luiken
ABSTRACT
In the insulin resistant heart, energy fuel selection shifts away from glucose utilization towards almost complete dependence on long-chain fatty acids (LCFA). This shift results in excessive cardiac lipid accumulation and eventually heart failure. Lipid-induced cardiomyopathy may be averted by strategies that increase glucose uptake without elevating LCFA uptake. Protein kinase-D1 (PKD1) is involved in contraction-induced glucose, but not LCFA, uptake allowing to hypothesize that this kinase is an attractive target to treat lipid-induced cardiomyopathy. For this, cardiospecific constitutively active PKD1 overexpression (caPKD1)-mice were subjected to an insulin resistance-inducing high fat-diet for 20-weeks. Substrate utilization was assessed by microPET and cardiac function by echocardiography. Cardiomyocytes were isolated for measurement of substrate uptake, lipid accumulation and insulin sensitivity. Wild-type mice on a high fat-diet displayed increased basal myocellular LCFA uptake, increased lipid deposition, greatly impaired insulin signaling, and loss of insulin-stimulated glucose and LCFA uptake, which was associated with concentric hypertrophic remodeling. The caPKD1 mice on high-fat diet showed none of these characteristics, whereas on low-fat diet a shift towards cardiac glucose utilization in combination with hypertrophy and ventricular dilation was observed. In conclusion, these data suggest that PKD pathway activation may be an attractive therapeutic strategy to mitigate lipid accumulation, insulin resistance and maladaptive remodeling in the lipid-overloaded heart, but this requires further investigation.
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
SAFC
BIS-TRIS
SAFC
BIS-TRIS
Sigma-Aldrich
Potassium carbonate, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium carbonate, meets USP testing specifications
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Potassium carbonate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium carbonate, anhydrous, free-flowing, −325 mesh, Redi-Dri, reagent grade, ≥98%
Sigma-Aldrich
Potassium carbonate, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Potassium carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Potassium carbonate, reagent grade, ≥98%, powder, −325 mesh
Sigma-Aldrich
Potassium carbonate, puriss., meets analytical specification of Ph. Helv., anhydrous, granulated, 99-101% (calc. to the dried substance)