Merck
  • Synthesis and biological evaluation of new pyridone-annelated isoindigos as anti-proliferative agents.
Synthesis and biological evaluation of new pyridone-annelated isoindigos as anti-proliferative agents.
Molecules (Basel, Switzerland) (2014-08-27)
Ayman M Saleh, Randa M Al-As'ad, Mustafa M El-Abadelah, Salim S Sabri, Jalal A Zahra, Ahmed S Alaskar, Ahmad Aljada
ABSTRACT
A selected set of substituted pyridone-annelated isoindigos 3a-f has been synthesized via interaction of 5- and 6-substituted oxindoles 2a-f with 6-ethyl-1,2,9-trioxopyrrolo[3,2-f]quinoline-8-carboxylic acid (1) in acetic acid at reflux. Among these isoindigos, the 5'-chloro and 5'-bromo derivatives 3b and 3d show strong and selective antiproliferative activities against a panel of human hematological and solid tumor cell-lines, but not against noncancerous cells, suggesting their potential use as anticancer agents. In all the tested cell lines, compound 3b was a 25%-50% more potent inhibitor of cell growth than 3d, suggesting the critical role of the substitution at 5'-position of the benzo-ring E. The IC50 values after 48 hours incubation with the 5'-chloro compound 3b were 6.60 µM in K562, 8.21 µM in THP-1, 8.97 µM in HepG2, 11.94 µM in MCF-7 and 14.59 µM in Caco-2 cancer cells, while the IC50 values in noncancerous HEK-293 and L-929 were 30.65 µM and 40.40 µM, respectively. In addition, compound 3b induced higher levels apoptosis in K562 cells than 3d, as determined by annexin V/7-AAD flowcytometry analysis. Therefore, further characterization of the antitproliferative mechanisms of compounds 3b and 3d may provide a novel chemotherapeutic agents.
Sigma-Aldrich
Hydroxylamine hydrochloride, JIS special grade, ≥98.0%
Sigma-Aldrich
Tin(II) chloride, anhydrous, powder, ≥99.99% trace metals basis
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydroxylamine hydrochloride, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.995% trace metals basis
Sigma-Aldrich
Hydroxylamine hydrochloride, ReagentPlus®, 99%
Sigma-Aldrich
Hydroxylamine hydrochloride, ACS reagent, 98.0%
Sigma-Aldrich
Tin(II) chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Tin(II) chloride, reagent grade, 98%
Sigma-Aldrich
Chloral hydrate, SAJ special grade, ≥99.7%
Sigma-Aldrich
Chloral hydrate, SAJ first grade, ≥99.5%
Sigma-Aldrich
Diphenyl ether, SAJ first grade, ≥98.0%
Supelco
4-Nitroaniline, analytical standard
Sigma-Aldrich
5-Chloro-2-oxindole, 98%
Sigma-Aldrich
Chloral hydrate
Sigma-Aldrich
6-Chloro-2-oxindole, 97%
Supelco
Diphenyl ether, for ion-selective electrodes, Selectophore, ≥99.9%
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Chloral hydrate, ≥99%
Sigma-Aldrich
4-Nitroaniline, ≥99%
Sigma-Aldrich
2-Oxindole, 97%
Sigma-Aldrich
Diphenyl ether, ≥99%, FG
Sigma-Aldrich
Diphenyl ether, ReagentPlus®, 99%
Sigma-Aldrich
Diphenyl ether, ReagentPlus®, ≥99%
Sigma-Aldrich
Iodoethane, contains copper as stabilizer, ReagentPlus®, 99%
Etoposide, European Pharmacopoeia (EP) Reference Standard