Merck
  • Formation of monodisperse hierarchical lipid particles utilizing microfluidic droplets in a nonequilibrium state.
Formation of monodisperse hierarchical lipid particles utilizing microfluidic droplets in a nonequilibrium state.
Langmuir : the ACS journal of surfaces and colloids (2015-02-12)
Masahiro Mizuno, Taro Toyota, Miki Konishi, Yoshiyuki Kageyama, Masumi Yamada, Minoru Seki
ABSTRACT
A new microfluidic process was used to generate unique micrometer-sized hierarchical lipid particles having spherical lipid-core and multilamellar-shell structures. The process includes three steps: (1) formation of monodisperse droplets in a nonequilibrium state at a microchannel confluence, using a phospholipid-containing water-soluble organic solvent as the dispersed phase and water as the continuous phase; (2) dissolution of the organic solvent of the droplet into the continuous phase and concentration of the lipid molecules; and (3) reconstitution of multilamellar lipid membranes and simultaneous formation of a lipid core. We demonstrated control of the lipid particle size by the process conditions and characterized the obtained particles by transmission electron microscopy and microbeam small-angle X-ray scattering analysis. In addition, we prepared various types of core-shell and core-core-shell particles incorporating hydrophobic/hydrophilic compounds, showing the applicability of the presented process to the production of drug-encapsulating lipid particles.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Diethyl ether, for residue analysis, JIS 5000
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Diethyl ether, JIS special grade, ≥99.5%
Sigma-Aldrich
Diethyl ether, SAJ first grade, ≥99.0%
Sigma-Aldrich
Diethyl ether, JIS 300, ≥99.5%, for residue analysis
Supelco
Rhodamine B, analytical standard
Supelco
Pyrene, analytical standard
Supelco
Pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Diethyl ether, analytical standard
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Rhodamine B solution, 0.2% in isopropanol, for TLC derivatization
Pyrene, BCR®, certified reference material
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Diethyl ether, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets