Merck
Length-dependent conductance of oligothiophenes.
Journal of the American Chemical Society (2014-07-09)
Brian Capozzi, Emma J Dell, Timothy C Berkelbach, David R Reichman, Latha Venkataraman, Luis M Campos
ABSTRACT
We have measured the single-molecule conductance of a family of oligothiophenes comprising 1-6 thiophene moieties terminated with methyl-sulfide linkers using the scanning tunneling microscope-based break-junction technique. We find an anomalous behavior: the peak of the conductance histogram distribution does not follow a clear exponential decay with increasing number of thiophene units in the chain. The electronic properties of the materials were characterized by optical spectroscopy and electrochemistry to gain an understanding of the factors affecting the conductance of these molecules. We postulate that different conformers in the junction are a contributing factor to the anomalous trend in the observed conductance as a function of molecule length.
Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
3-Hexylthiophene, ≥99%
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
2,2′-Bithiophene, 99%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Triethylamine, analytical standard
Sigma-Aldrich
Thiophene, ≥99%
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
2,2′:5′,2′′-Terthiophene, 99%
Supelco
Thiophene, analytical standard
Sigma-Aldrich
Potassium carbonate, meets USP testing specifications
Sigma-Aldrich
Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Potassium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium carbonate, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Potassium carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Potassium carbonate, anhydrous, free-flowing, −325 mesh, Redi-Dri, reagent grade, ≥98%
Sigma-Aldrich
Potassium carbonate, meets analytical specification of Ph. Helv., puriss., anhydrous, granulated, 99-101% (calc. to the dried substance)
Sigma-Aldrich
Potassium carbonate, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium carbonate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium carbonate, reagent grade, ≥98%, powder, −325 mesh