Merck
  • Pharmacokinetics of omeprazole and its metabolites in three phases of menstrual cycle.
Pharmacokinetics of omeprazole and its metabolites in three phases of menstrual cycle.
European journal of drug metabolism and pharmacokinetics (2014-01-07)
Shabnam Nazir, Zafar Iqbal, Lateef Ahmad, Yasar Shah, Fazli Nasir
ABSTRACT
Omeprazole (OMP) is effective in the treatment of gastric hyperacidity and is metabolized by CYP2C19 and CYP3A4. These enzymes are modulated by estrogen and progesterone which regulate the menstrual cycle. The variations in the pharmacokinetics (PK) of many drugs like amphetamine, benzodiazepines and caffeine have been reported during menstrual cycle. In present study, the PK of the omeprazole and its metabolites was investigated during various phases of the menstrual cycle. A single oral dose, open-label, non-controlled, pharmacokinetic study of omeprazole was conducted in healthy young/premenopausal females (n = 16). The PK of omeprazole, 5-hydroxy-omeprazole and omeprazole sulphone was evaluated in three phases of menstrual cycle. The blood samples were analyzed using reversed-phase HPLC coupled with UV detector and the PK data were evaluated. The activities of CYP2C19 and CYP3A4 were determined as AUC(OH-OMP)/AUC(OMP) and AUC(OMP-SUL)/AUC(OMP), respectively. Omeprazole showed significantly (p < 0.05) higher [Formula: see text] and CL/F in follicular and menstrual phases, respectively. The [Formula: see text] of 5-hydroxy omeprazole was also significantly (p < 0.05) higher in follicular phase. The metabolic ratios (MR) of 5-hydroxy omeprazole and omeprazole sulphone were lower in follicular phase compared with the luteal phase. The present study suggests that high estrogen levels of follicular phase may result in increased absorption of omeprazole. The lower MR for 5-hydroxy omeprazole and omeprazole sulphone in follicular phase as compared to luteal phase suggests that metabolism of omeprazole is low in follicular phase as compared to luteal phase, which is progesterone-dominant phase. However, the clinical significance for these findings needs to be determined.
Product Description
Supelco
5-Hydroxyomeprazole, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
USP
Omeprazole Related Compound A, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Potassium phosphate monobasic, ≥99.5%
Supelco
Omeprazole, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Omeprazole sulfone, analytical standard
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)