Merck
  • In-vivo evaluation of human recombinant Co-arginase against A375 melanoma xenografts.
In-vivo evaluation of human recombinant Co-arginase against A375 melanoma xenografts.
Melanoma research (2014-10-12)
Vaidehi Agrawal, Jung Hee Woo, Jeremy P Mauldin, Everett M Stone, Cynthia J Meininger, Chanhee Jo, Keri Kleypas, Eugene P Frenkel, Arthur E Frankel
ABSTRACT
Metastatic melanoma is a deadly form of cancer with few therapeutic options and the cause of more than 9480 deaths annually in the USA alone. Novel treatment options for this disease are urgently needed. Here we test the efficacy of a novel melanoma drug, the human recombinant Co-arginase (CoArgIPEG), against an aggressive A375 melanoma mouse model. CoArgIPEG is a modification of the naturally occurring human enzyme with improved stability, catalytic activity, and potentially lower immunogenicity compared with current amino acid-depleting drugs. Marked tumor growth reductions (mean P=0.0057) with apoptosis induction and proliferation inhibition are noted with CoArgIPEG treatment, both in the presence and in the absence of supplemental citrulline. Further, improved therapeutic efficacy has been noted against A375 xenografts relative to the naturally occurring human recombinant arginase enzyme at lower doses of CoArgIPEG. Unfortunately, after 1 month, half of the relapsing tumors showed argininosuccinate synthase induction, which correlated with Ser62-phosphorylated cMyc. Although argininosuccinate synthase induction could not be induced in vitro, a drug targeting pathway previously demonstrated to be associated with Ser62 cMyc phosphorylation - U0126 - in combination with CoArgIPEG demonstrated an in-vitro synergistic response (combination indices 0.13±0.10 and 0.14±0.10 with or without citrulline, respectively). Overall, favorable efficacy and potential synergy with other antimelanoma drugs support CoArgIPEG as a potent, novel cancer therapeutic.
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (titration)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, suitable for plant cell culture
SAFC
Glycine
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
BIS-TRIS
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
SAFC
MOPS
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
SAFC
BIS-TRIS