Merck
  • Evidence for organic cation transporter-mediated metformin transport and 5'-adenosine monophosphate-activated protein kinase activation in rat skeletal muscles.
Evidence for organic cation transporter-mediated metformin transport and 5'-adenosine monophosphate-activated protein kinase activation in rat skeletal muscles.
Metabolism: clinical and experimental (2014-12-01)
Rieko Oshima, Mayumi Yamada, Eriko Kurogi, Yohei Ogino, Yasuhiro Serizawa, Satoshi Tsuda, Xiao Ma, Tatsuro Egawa, Tatsuya Hayashi
ABSTRACT
5'-Adenosine monophosphate-activated protein kinase (AMPK) is a key molecule of metabolic enhancement in skeletal muscle. We investigated whether metformin (MET) acts directly on skeletal muscle, is transported into skeletal muscle via organic cation transporters (OCTs), and activates AMPK. Isolated rat epitrochlearis and soleus muscles were incubated in vitro either in the absence or in the presence of MET. The activation status of AMPK, the intracellular energy status, and glucose and MET transport activity were then evaluated. The effect of cimetidine, which is an OCT inhibitor, on AMPK activation was also examined. MET (10 mmol/L, ≥60 min) increased the phosphorylation of Thr¹⁷² at the catalytic α subunit of AMPK in both muscles. AMPK activity assays showed that both AMPKα1 and AMPKα2 activity increased significantly. The AMPK activation was associated with energy deprivation, which was estimated from the ATP, phosphocreatine (PCr), and glycogen content, and with increased rates of 3-O-methyl-D-glucose (3MG) transport. MET did not change the basal phosphorylation status of insulin receptor signaling molecules. MET was transported into the cytoplasm in a time-dependent manner, and cimetidine suppressed MET-induced AMPK phosphorylation and 3MG transport. These results suggest that MET is acutely transported into skeletal muscle by OCTs, and stimulates AMPKα1 and α2 activity in both fast- and slow-twitch muscle types, at least in part by reducing the energy state.
Product Description
Cimetidine for peak identification, European Pharmacopoeia (EP) Reference Standard
Cimetidine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Cimetidine, Pharmaceutical Secondary Standard; Certified Reference Material
Cimetidine hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Cimetidine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cimetidine
Cimetidine, European Pharmacopoeia (EP) Reference Standard