Merck
  • Cross-inhibition of NMBR and GRPR signaling maintains normal histaminergic itch transmission.
Cross-inhibition of NMBR and GRPR signaling maintains normal histaminergic itch transmission.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-09-12)
Zhong-Qiu Zhao, Li Wan, Xian-Yu Liu, Fu-Quan Huo, Hui Li, Devin M Barry, Stephanie Krieger, Seungil Kim, Zhong-Chun Liu, Jinbin Xu, Buck E Rogers, Yun-Qing Li, Zhou-Feng Chen
ABSTRACT
We previously showed that gastrin-releasing peptide receptor (GRPR) in the spinal cord is important for mediating nonhistaminergic itch. Neuromedin B receptor (NMBR), the second member of the mammalian bombesin receptor family, is expressed in a largely nonoverlapping pattern with GRPR in the superficial spinal cord, and its role in itch transmission remains unclear. Here, we report that Nmbr knock-out (KO) mice exhibited normal scratching behavior in response to intradermal injection of pruritogens. However, mice lacking both Nmbr and Grpr (DKO mice) showed significant deficits in histaminergic itch. In contrast, the chloroquine (CQ)-evoked scratching behavior of DKO mice is not further reduced compared with Grpr KO mice. These results suggest that NMBR and GRPR could compensate for the loss of each other to maintain normal histamine-evoked itch, whereas GRPR is exclusively required for CQ-evoked scratching behavior. Interestingly, GRPR activity is enhanced in Nmbr KO mice despite the lack of upregulation of Grpr expression; so is NMBR in Grpr KO mice. We found that NMB acts exclusively through NMBR for itch transmission, whereas GRP can signal through both receptors, albeit to NMBR to a much lesser extent. Although NMBR and NMBR(+) neurons are dispensable for histaminergic itch, GRPR(+) neurons are likely to act downstream of NMBR(+) neurons to integrate NMB-NMBR-encoded histaminergic itch information in normal physiological conditions. Together, we define the respective function of NMBR and GRPR in itch transmission, and reveal an unexpected relationship not only between the two receptors but also between the two populations of interneurons in itch signaling.
Sigma-Aldrich
Capsaicin, from Capsicum sp., ≥50% (HPLC)
Sigma-Aldrich
Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia), Isolectin B4 (BSI-B4), FITC conjugate, lyophilized powder
Supelco
Capsaicin, analytical standard
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
Capsaicin, natural
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
Anti-Substance P Receptor Antibody, pain, serum, Chemicon®
Supelco
Histamine, analytical standard
USP
Capsaicin, United States Pharmacopeia (USP) Reference Standard
Supelco
Capsaicin, Pharmaceutical Secondary Standard; Certified Reference Material
Capsaicin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Serotonin creatinine sulfate monohydrate, powder