Skip to Content
Merck
All Photos(1)

Key Documents

L2895

Sigma-Aldrich

Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia)

Isolectin B4 (BSI-B4), FITC conjugate, lyophilized powder

Synonym(s):

Bandeirea simplicifolia agglutinin, BS-I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

conjugate

FITC conjugate

Quality Level

form

lyophilized powder

potency

<50 μg per mL agglutination activity (using human blood group B erythrocytes)

composition

Protein, ≥70% Lowry (balance sodium citrate)

extent of labeling

≥1 mol FITC per mol lectin

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Lectins are carbohydrate-binding proteins, omnipresent, found in fungi, plants and animals. The structure of lectin is diversely studied in plants and animals. The secondary structure of this protein is rich in β-strands and possesses carbohydrate binding sites on the surface.

Application

Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia) has been used for microglia detection in murine astrocytes culture by staining the culture with lectin BS1-B4. It has also been used in the preparation of staining solution to incubate the tissue sections.

Biochem/physiol Actions

BS-I has a major affinity for terminal α-D-galactosyl residues with a secondary affinity for terminal N-acetyl-α-D-galactosaminyl residues.
Lectin is known to be useful in glycoconjugate characterizing, imaging and targeting. Its use in a microarray assay, enable efficient glycome profiling, because of its specific interaction with oligosaccharides, glycoproteins and glycolipids. In plants and fungi, lectin defends against pathogens/feeders. Lectin participates in host recognition and tissue adhesion, thereby aids in the pathogenesis of microorganism.

Packaging

Package size based on protein content by Lowry.

Other Notes

BS-I is a tetrameric lectin consisting of two types of subunits designated A and B. There are five BS-I isolectins with different subunit composition: BSI-B4, BSI-AB3, BSI-A2B2, BSI-A3B and BSI-A4. BSI-B4 is blood group B specific and has an exclusive affinity for terminal α-D-galactosyl residues, whereas BSI-A4 has blood group A specificity and has a major affinity for terminal N-acetyl-α-D-galactosaminyl residues.

Analysis Note

Agglutination activity is expressed in μg/mL and is determined from serial dilutions of a 1 mg/mL solution using phosphate buffered saline, pH 6.8, containing, for each lectin, calcium, magnesium, and manganese at different concentrations. This activity is the lowest concentration to agglutinate a 2% suspension of appropriate erythrocytes after 1 hr incubation at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduced adipose tissue oxygenation in human obesity: evidence for rarefaction, macrophage chemotaxis, and inflammation without an angiogenic response
Pasarica M, et al.
Diabetes, 58(3), 718-725 (2009)
Interleukin-18 involvement in hypoxic--ischemic brain injury
Hedtjarn M, et al.
The Journal of Neuroscience, 22(14), 5910-5919 (2002)
Gamma Interferon-Induced Inhibition ofToxoplasma gondii in Astrocytes Is Mediated by IGTP
Halonen SK, et al.
Infection and Immunity, 69(9), 5573-5576 (2001)
Evgeny Levin et al.
Scientific reports, 6, 38928-38928 (2016-12-14)
Delivery and expression of recombinant genes, a key methodology for many applications in biological research, remains a challenge especially for mature neurons. Here, we report easy, highly efficient and well tolerated transduction of adult peripheral and central neuronal populations of
K A Vessey et al.
The Journal of comparative neurology, 519(3), 506-527 (2010-12-31)
Retinal neovascularization, such as that occurring in proliferative diabetic retinopathy and retinopathy of prematurity, can have serious effects on visual function. By using a mouse model of neovascularization, oxygen-induced retinopathy (OIR), the interplay among angiogenesis, neuronal function, and the macro-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service