Merck
  • Risk of upper gastrointestinal bleeding from different drug combinations.
Risk of upper gastrointestinal bleeding from different drug combinations.
Gastroenterology (2014-06-18)
Gwen M C Masclee, Vera E Valkhoff, Preciosa M Coloma, Maria de Ridder, Silvana Romio, Martijn J Schuemie, Ron Herings, Rosa Gini, Giampiero Mazzaglia, Gino Picelli, Lorenza Scotti, Lars Pedersen, Ernst J Kuipers, Johan van der Lei, Miriam C J M Sturkenboom
ABSTRACT
Concomitant use of nonsteroidal anti-inflammatory drugs (NSAIDs) and low-dose aspirin increases the risk of upper gastrointestinal bleeding (UGIB). Guidelines suggest avoiding certain drug combinations, yet little is known about the magnitude of their interactions. We estimated the risk of UGIB during concomitant use of nonselective (ns)NSAIDs, cyclooxygenase -2 selective inhibitors (COX-2 inhibitors), and low-dose aspirin with other drugs. We performed a case series analysis of data from 114,835 patients with UGIB (930,888 person-years of follow-up) identified from 7 population-based health care databases (approximately 20 million subjects). Each patient served as his or her own control. Drug exposure was determined based on prescriptions of nsNSAIDs, COX-2 inhibitors, or low-dose aspirin, alone and in combination with other drugs that affect the risk of UGIB. We measured relative risk (incidence rate ratio [IRR] during drug exposure vs nonexposure) and excess risk due to concomitant drug exposure (relative excess risk due to interaction [RERI]). Monotherapy with nsNSAIDs increased the risk of diagnosis of UGIB (IRR, 4.3) to a greater extent than monotherapy with COX-2 inhibitors (IRR, 2.9) or low-dose aspirin (IRR, 3.1). Combination therapy generally increased the risk of UGIB; concomitant nsNSAID and corticosteroid therapies increased the IRR to the greatest extent (12.8) and also produced the greatest excess risk (RERI, 5.5). Concomitant use of nsNSAIDs and aldosterone antagonists produced an IRR for UGIB of 11.0 (RERI, 4.5). Excess risk from concomitant use of nsNSAIDs with selective serotonin reuptake inhibitors (SSRIs) was 1.6, whereas that from use of COX-2 inhibitors with SSRIs was 1.9 and that for use of low-dose aspirin with SSRIs was 0.5. Excess risk of concomitant use of nsNSAIDs with anticoagulants was 2.4, of COX-2 inhibitors with anticoagulants was 0.1, and of low-dose aspirin with anticoagulants was 1.9. Based on a case series analysis, concomitant use of nsNSAIDs, COX-2 inhibitors, or low-dose aspirin with SSRIs significantly increases the risk of UGIB. Concomitant use of nsNSAIDs or low-dose aspirin, but not COX-2 inhibitors, with corticosteroids, aldosterone antagonists, or anticoagulants produces significant excess risk of UGIB.
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Aspirin, meets USP testing specifications